missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dimethyl (2-oxopropyl)phosphonate, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 177611000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
for C-C Bond Formation (Olefination)Specifications
Dimethyl (2-oxopropyl)phosphonate | |
76°C to 79°C (3.0mmHg) | |
Authentic | |
Glass bottle | |
CH3COCH2P(O)(OCH3)2 | |
MFCD00008769 | |
dimethyl 2-oxopropyl phosphonate, dimethyl acetylmethylphosphonate, dimethyl 2-oxopropylphosphonate, dimethyl acetonylphosphonate, 2-oxopropyl phosphonic acid dimethyl ester, 1-dimethoxycarbonyl acetone, acmc-209jmw, dimethoxyphosphoryl acetone | |
CC(=O)CP(=O)(OC)OC | |
166.11 | |
166.11 |
4202-14-6 | |
93°C | |
95% | |
C5H11O4P | |
1.4380 to 1.441 | |
100g | |
UOWIYNWMROWVDG-UHFFFAOYSA-N | |
1-dimethoxyphosphorylpropan-2-one | |
77872 | |
95% |
Chemical Identifiers
4202-14-6 | |
166.11 | |
UOWIYNWMROWVDG-UHFFFAOYSA-N | |
77872 | |
CC(=O)CP(=O)(OC)OC |
C5H11O4P | |
MFCD00008769 | |
dimethyl 2-oxopropyl phosphonate, dimethyl acetylmethylphosphonate, dimethyl 2-oxopropylphosphonate, dimethyl acetonylphosphonate, 2-oxopropyl phosphonic acid dimethyl ester, 1-dimethoxycarbonyl acetone, acmc-209jmw, dimethoxyphosphoryl acetone | |
1-dimethoxyphosphorylpropan-2-one |
Safety and Handling
EINECSNumber : 224-110-5
RUO â Research Use Only