Learn More
Dimethyl 5-aminoisophthalate, 98%, Thermo Scientific Chemicals
$71.01 - $166.58
Chemical Identifiers
CAS | 99-27-4 |
---|---|
Molecular Formula | C10H11NO4 |
Molecular Weight (g/mol) | 209.201 |
MDL Number | MFCD00008435 |
InChI Key | DEKPYXUDJRABNK-UHFFFAOYSA-N |
PubChem CID | 66831 |
IUPAC Name | dimethyl 5-aminobenzene-1,3-dicarboxylate |
SMILES | COC(=O)C1=CC(=CC(=C1)N)C(=O)OC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2076114
|
Thermo Scientific Chemicals
B2076114 |
25 g |
N/A
|
|
|||||
AAB2076122
|
Thermo Scientific Chemicals
B2076122 |
100 g |
N/A
|
|
|||||
Description
Dimethyl 5-aminoisophthalate is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. Hexagonal assembly of double helices of luminescent calcium 5-aminoisophthalate coordination polymer leaves enough room for the formation of loosely held water pipes, which could be dehydrated at very low temperatures.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDimethyl 5-aminoisophthalate is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. Hexagonal assembly of double helices of luminescent calcium 5-aminoisophthalate coordination polymer leaves enough room for the formation of loosely held water pipes, which could be dehydrated at very low temperatures.
Solubility
Soluble in Chloroform.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Chemical Identifiers
99-27-4 | |
209.201 | |
DEKPYXUDJRABNK-UHFFFAOYSA-N | |
dimethyl 5-aminobenzene-1,3-dicarboxylate |
C10H11NO4 | |
MFCD00008435 | |
66831 | |
COC(=O)C1=CC(=CC(=C1)N)C(=O)OC |
Specifications
99-27-4 | |
C10H11NO4 | |
25 g | |
dimethyl 5-aminoisophthalate, 3,5-dicarbomethoxyaniline, 5-aminoisophthalic acid dimethyl ester, 1,3-benzenedicarboxylic acid, 5-amino-, dimethyl ester, 1,3-dimethyl 5-aminobenzene-1,3-dicarboxylate, dimethyl5-aminoisophthalate, 5-amino-isophthalic acid dimethyl ester, 5-amino-dimethyl iso-phthalate, 5-amino-dimethylisophthalate, dimethyl-5-aminoisophthalate | |
DEKPYXUDJRABNK-UHFFFAOYSA-N | |
dimethyl 5-aminobenzene-1,3-dicarboxylate | |
66831 | |
98% |
178°C to 182°C | |
MFCD00008435 | |
2116565 | |
Soluble in Chloroform. | |
COC(=O)C1=CC(=CC(=C1)N)C(=O)OC | |
209.201 | |
209.2 | |
Dimethyl 5-aminoisophthalate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 202-743-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only