missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dimethyl 5-Aminoisophthalate 98.0+%, TCI America™
Supplier: TCI America A1586250G
Specifications
Dimethyl 5-Aminoisophthalate | |
181°C | |
MFCD00008435 | |
dimethyl 5-aminoisophthalate, 3,5-dicarbomethoxyaniline, 5-aminoisophthalic acid dimethyl ester, 1,3-benzenedicarboxylic acid, 5-amino-, dimethyl ester, 1,3-dimethyl 5-aminobenzene-1,3-dicarboxylate, dimethyl5-aminoisophthalate, 5-amino-isophthalic acid dimethyl ester, 5-amino-dimethyl iso-phthalate, 5-amino-dimethylisophthalate, dimethyl-5-aminoisophthalate | |
COC(=O)C1=CC(=CC(=C1)N)C(=O)OC | |
209.201 | |
209.20 | |
Crystalline Powder |
99-27-4 | |
C10H11NO4 | |
250 g | |
DEKPYXUDJRABNK-UHFFFAOYSA-N | |
dimethyl 5-aminobenzene-1,3-dicarboxylate | |
66831 | |
≥98.0% (HPLC,T) |
Chemical Identifiers
99-27-4 | |
209.201 | |
DEKPYXUDJRABNK-UHFFFAOYSA-N | |
dimethyl 5-aminobenzene-1,3-dicarboxylate |
C10H11NO4 | |
MFCD00008435 | |
66831 | |
COC(=O)C1=CC(=CC(=C1)N)C(=O)OC |
Safety and Handling
EINECSNumber : (3)-3026
TSCA : Yes