missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethyl 5-bromoisophthalate, 98%, Thermo Scientific Chemicals

$63.89 - $286.08

Chemical Identifiers

CAS 51760-21-5
Molecular Formula C10H9BrO4
Molecular Weight (g/mol) 273.082
MDL Number MFCD00078709
InChI Key QUJINGKSNJNXEB-UHFFFAOYSA-N
Synonym dimethyl 5-bromoisophthalate, 5-bromoisophthalic acid dimethyl ester, dimethyl-5-bromoisophthalate, 1,3-benzenedicarboxylic acid, 5-bromo-, dimethyl ester, 1,3-dimethyl 5-bromobenzene-1,3-dicarboxylate, dimethyl5-bromoisophthalate, 5-bromo-isophthalic acid dimethyl ester, 1,3-benzenedicarboxylic acid, 5-bromo-, 1,3-dimethyl ester, methyl 5-bromo-3-methoxycarbonyl benzoate, 5-bromoisophthalic acid, dimethyl ester
PubChem CID 103954
IUPAC Name dimethyl 5-bromobenzene-1,3-dicarboxylate
SMILES COC(=O)C1=CC(=CC(=C1)Br)C(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5062903
View Documents
Thermo Scientific Chemicals
H5062903
1 g
Each for $63.89
 
AAH5062906
View Documents
Thermo Scientific Chemicals
H5062906
5 g
Each for $286.08
 
Description

Description

Dimethyl 5-bromoisophthalate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl 5-bromoisophthalate is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

51760-21-5
273.082
QUJINGKSNJNXEB-UHFFFAOYSA-N
103954
COC(=O)C1=CC(=CC(=C1)Br)C(=O)OC
C10H9BrO4
MFCD00078709
dimethyl 5-bromoisophthalate, 5-bromoisophthalic acid dimethyl ester, dimethyl-5-bromoisophthalate, 1,3-benzenedicarboxylic acid, 5-bromo-, dimethyl ester, 1,3-dimethyl 5-bromobenzene-1,3-dicarboxylate, dimethyl5-bromoisophthalate, 5-bromo-isophthalic acid dimethyl ester, 1,3-benzenedicarboxylic acid, 5-bromo-, 1,3-dimethyl ester, methyl 5-bromo-3-methoxycarbonyl benzoate, 5-bromoisophthalic acid, dimethyl ester
dimethyl 5-bromobenzene-1,3-dicarboxylate

Specifications

51760-21-5
C10H9BrO4
1 g
Insoluble in water.
COC(=O)C1=CC(=CC(=C1)Br)C(=O)OC
273.082
273.08
Dimethyl 5-bromoisophthalate
88°C to 90°C
MFCD00078709
dimethyl 5-bromoisophthalate, 5-bromoisophthalic acid dimethyl ester, dimethyl-5-bromoisophthalate, 1,3-benzenedicarboxylic acid, 5-bromo-, dimethyl ester, 1,3-dimethyl 5-bromobenzene-1,3-dicarboxylate, dimethyl5-bromoisophthalate, 5-bromo-isophthalic acid dimethyl ester, 1,3-benzenedicarboxylic acid, 5-bromo-, 1,3-dimethyl ester, methyl 5-bromo-3-methoxycarbonyl benzoate, 5-bromoisophthalic acid, dimethyl ester
QUJINGKSNJNXEB-UHFFFAOYSA-N
dimethyl 5-bromobenzene-1,3-dicarboxylate
103954
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 257-386-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only