missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethyl allylphosphonate, tech. 85%, Thermo Scientific Chemicals

$211.84 - $211.84

Chemical Identifiers

CAS 757-54-0
Molecular Formula C5H11O3P
Molecular Weight (g/mol) 150.114
MDL Number MFCD00014951
InChI Key ZOSQAGGCVFVCNO-UHFFFAOYSA-N
Synonym dimethyl allylphosphonate, allyl phosphonic acid dimethyl ester, dimethyl prop-2-en-1-ylphosphonate, acmc-20al6z, dimethyl allyl phosphonate, allylphosphonic acid dimethyl, allylphosphonic acid dimethyl ester
PubChem CID 136570
IUPAC Name 3-dimethoxyphosphorylprop-1-ene
SMILES COP(=O)(CC=C)OC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1056706
View Documents
Thermo Scientific Chemicals
L1056706
5 g
Each for $211.84
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

757-54-0
150.114
ZOSQAGGCVFVCNO-UHFFFAOYSA-N
136570
COP(=O)(CC=C)OC
C5H11O3P
MFCD00014951
dimethyl allylphosphonate, allyl phosphonic acid dimethyl ester, dimethyl prop-2-en-1-ylphosphonate, acmc-20al6z, dimethyl allyl phosphonate, allylphosphonic acid dimethyl, allylphosphonic acid dimethyl ester
3-dimethoxyphosphorylprop-1-ene

Specifications

757-54-0
C5H11O3P
MFCD00014951
1756526
ZOSQAGGCVFVCNO-UHFFFAOYSA-N
3-dimethoxyphosphorylprop-1-ene
136570
85%
Dimethyl allylphosphonate
41°C to 42°C (0.5 mmHg)
1.437
5 g
dimethyl allylphosphonate, allyl phosphonic acid dimethyl ester, dimethyl prop-2-en-1-ylphosphonate, acmc-20al6z, dimethyl allyl phosphonate, allylphosphonic acid dimethyl, allylphosphonic acid dimethyl ester
COP(=O)(CC=C)OC
150.114
150.12
Technical
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only