missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethyl aminomalonate hydrochloride, 97%, Thermo Scientific™

Chemical Identifiers

CAS 16115-80-3
Molecular Formula C5H10ClNO4
Molecular Weight (g/mol) 183.59
MDL Number MFCD00038880
InChI Key QWNDKNJSEWOEDM-UHFFFAOYSA-N
Synonym dimethyl 2-aminomalonate hydrochloride, dimethyl aminomalonate hydrochloride, dimethyl aminomalonate hcl, aminomalonic acid dimethyl ester hydrochloride, 1,3-dimethyl 2-aminopropanedioate hydrochloride, dimethyl 2-aminopropanedioate hydrochloride, acmc-1bp9z, 2-amino-malonic acid dimethyl ester hydrochloride, 2-aminomalonic acid dimethyl ester hydrochloride
PubChem CID 2734846
IUPAC Name dimethyl 2-aminopropanedioate;hydrochloride
SMILES [H+].[Cl-].COC(=O)C(N)C(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC374970050
SDS View Documents
Thermo Scientific Chemicals
374970050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC374970250
SDS View Documents
Thermo Scientific Chemicals
374970250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

16115-80-3
183.59
QWNDKNJSEWOEDM-UHFFFAOYSA-N
2734846
[H+].[Cl-].COC(=O)C(N)C(=O)OC
C5H10ClNO4
MFCD00038880
dimethyl 2-aminomalonate hydrochloride, dimethyl aminomalonate hydrochloride, dimethyl aminomalonate hcl, aminomalonic acid dimethyl ester hydrochloride, 1,3-dimethyl 2-aminopropanedioate hydrochloride, dimethyl 2-aminopropanedioate hydrochloride, acmc-1bp9z, 2-amino-malonic acid dimethyl ester hydrochloride, 2-aminomalonic acid dimethyl ester hydrochloride
dimethyl 2-aminopropanedioate;hydrochloride

Specifications

16115-80-3
96% min. (Argentometry)
C5H10ClNO4
MFCD00038880
dimethyl 2-aminomalonate hydrochloride, dimethyl aminomalonate hydrochloride, dimethyl aminomalonate hcl, aminomalonic acid dimethyl ester hydrochloride, 1,3-dimethyl 2-aminopropanedioate hydrochloride, dimethyl 2-aminopropanedioate hydrochloride, acmc-1bp9z, 2-amino-malonic acid dimethyl ester hydrochloride, 2-aminomalonic acid dimethyl ester hydrochloride
[H+].[Cl-].COC(=O)C(N)C(=O)OC
183.59
183.59
Dimethyl aminomalonate hydrochloride
Authentic
Glass bottle
H2NCH(CO2CH3)2·HCl
5g
QWNDKNJSEWOEDM-UHFFFAOYSA-N
dimethyl 2-aminopropanedioate;hydrochloride
2734846
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 240-284-5

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only