missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dimethyl Benzylmalonate 95.0+%, TCI America™
$152.08 - $454.25
Chemical Identifiers
CAS | 49769-78-0 |
---|---|
Molecular Formula | C12H14O4 |
Molecular Weight (g/mol) | 222.24 |
MDL Number | MFCD01445471 |
InChI Key | ZMYJSOIMFGAQRQ-UHFFFAOYSA-N |
Synonym | Benzylmalonic Acid Dimethyl Ester |
PubChem CID | 10900856 |
IUPAC Name | 1,3-dimethyl 2-benzylpropanedioate |
SMILES | COC(=O)C(CC1=CC=CC=C1)C(=O)OC |
Chemical Identifiers
49769-78-0 | |
222.24 | |
ZMYJSOIMFGAQRQ-UHFFFAOYSA-N | |
10900856 | |
COC(=O)C(CC1=CC=CC=C1)C(=O)OC |
C12H14O4 | |
MFCD01445471 | |
Benzylmalonic Acid Dimethyl Ester | |
1,3-dimethyl 2-benzylpropanedioate |
Specifications
49769-78-0 | |
285°C | |
MFCD01445471 | |
Benzylmalonic Acid Dimethyl Ester | |
COC(=O)C(CC1=CC=CC=C1)C(=O)OC | |
222.24 | |
222.24 | |
Liquid |
Colorless | |
C12H14O4 | |
5 g | |
ZMYJSOIMFGAQRQ-UHFFFAOYSA-N | |
1,3-dimethyl 2-benzylpropanedioate | |
10900856 | |
≥95.0% (GC) | |
Dimethyl Benzylmalonate |
Safety and Handling
TSCA : No