missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethyl carbonate, 99+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals

$114.98 - $285.92

Chemical Identifiers

CAS 616-38-6
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
MDL Number MFCD00008420
InChI Key IEJIGPNLZYLLBP-UHFFFAOYSA-N
Synonym methyl carbonate, carbonic acid, dimethyl ester, carbonic acid dimethyl ester, methyl carbonate meo 2co, unii-ke9j097spn, dimethylcarbonate, ch3ocooch3, ke9j097spn, dimethyl ester of carbonic acid, dsstox_cid_9192
PubChem CID 12021
ChEBI CHEBI:36596
IUPAC Name dimethyl carbonate
SMILES COC(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC428731000
SDS View Documents
Thermo Scientific Chemicals
428731000
100 mL AcroSeal™ Glass Bottle
Each for $114.98
 
AC428730010
SDS View Documents
Thermo Scientific Chemicals
428730010
1 L AcroSeal™ Glass Bottle
Each for $285.92
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications

Chemical Identifiers

616-38-6
90.08
IEJIGPNLZYLLBP-UHFFFAOYSA-N
12021
dimethyl carbonate
C3H6O3
MFCD00008420
methyl carbonate, carbonic acid, dimethyl ester, carbonic acid dimethyl ester, methyl carbonate meo 2co, unii-ke9j097spn, dimethylcarbonate, ch3ocooch3, ke9j097spn, dimethyl ester of carbonic acid, dsstox_cid_9192
CHEBI:36596
COC(=O)OC

Specifications

616-38-6
Brown to White
90.0°C
Authentic
AcroSeal™ Glass Bottle
1.3672 to 1.3692
MFCD00008420
1.069
methyl carbonate, carbonic acid, dimethyl ester, carbonic acid dimethyl ester, methyl carbonate meo 2co, unii-ke9j097spn, dimethylcarbonate, ch3ocooch3, ke9j097spn, dimethyl ester of carbonic acid, dsstox_cid_9192
IEJIGPNLZYLLBP-UHFFFAOYSA-N
dimethyl carbonate
12021
CHEBI:36596
99+%
Crystalline Powder
2.0°C to 4.0°C
1.0690g/mL
18°C
99% min. (GC)
C3H6O3
(CH3O)2CO
100 mL
15, 3263
Solubility in water: 139g/L. Other solubilities: soluble in all proportions with organic alcohols,soluble in all proportions with esters and ketones,miscible with acids and alkalies
COC(=O)OC
90.08
0.625 mPa.s (20°C)
90.08
Extra Dry
Dimethyl carbonate, Extra Dry
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 210-478-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only