missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Citraconate 99.0+%, TCI America™
$158.44 - $417.56
Chemical Identifiers
| CAS | 617-54-9 |
|---|---|
| Molecular Formula | C7H10O4 |
| Molecular Weight (g/mol) | 158.15 |
| MDL Number | MFCD00059306 |
| InChI Key | WQEXBUQDXKPVHR-SNAWJCMRSA-N |
| Synonym | Citraconic Acid Dimethyl Ester |
| PubChem CID | 5355715 |
| IUPAC Name | 1,4-dimethyl (2E)-2-methylbut-2-enedioate |
| SMILES | COC(=O)\C=C(/C)C(=O)OC |
Chemical Identifiers
| 617-54-9 | |
| 158.15 | |
| WQEXBUQDXKPVHR-SNAWJCMRSA-N | |
| 5355715 | |
| COC(=O)\C=C(/C)C(=O)OC |
| C7H10O4 | |
| MFCD00059306 | |
| Citraconic Acid Dimethyl Ester | |
| 1,4-dimethyl (2E)-2-methylbut-2-enedioate |
Specifications
| 617-54-9 | |
| 211°C | |
| MFCD00059306 | |
| Citraconic Acid Dimethyl Ester | |
| COC(=O)\C=C(/C)C(=O)OC | |
| 158.15 | |
| 158.15 | |
| Liquid |
| Yellow | |
| C7H10O4 | |
| 5 g | |
| WQEXBUQDXKPVHR-SNAWJCMRSA-N | |
| 1,4-dimethyl (2E)-2-methylbut-2-enedioate | |
| 5355715 | |
| ≥99.0% (GC) | |
| Dimethyl Citraconate |
Safety and Handling
RTECSNumber : GE6700000
TSCA : Yes