Learn More
Dimethyl diselenide, 99%, Thermo Scientific Chemicals
$164.78 - $526.04
Chemical Identifiers
CAS | 7101-31-7 |
---|---|
Molecular Formula | C2H6Se2 |
Molecular Weight (g/mol) | 188.012 |
MDL Number | MFCD00008254 |
InChI Key | VLXBWPOEOIIREY-UHFFFAOYSA-N |
Synonym | dimethyl diselenide, dimethyldiselenide, methyl diselenide, diselenide, dimethyl, ch3se 2, methyldiselanyl methane, diselenobismethane, 1,2-dimethyldiselane, acmc-209ogy, methyl diselenide, 8ci |
PubChem CID | 23496 |
IUPAC Name | (methyldiselanyl)methane |
SMILES | C[Se][Se]C |
Description
Dimethyl diselenide is a powerful reagent used for identification of distonic radical cations
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7101-31-7 | |
188.012 | |
VLXBWPOEOIIREY-UHFFFAOYSA-N | |
23496 | |
C[Se][Se]C |
C2H6Se2 | |
MFCD00008254 | |
dimethyl diselenide, dimethyldiselenide, methyl diselenide, diselenide, dimethyl, ch3se 2, methyldiselanyl methane, diselenobismethane, 1,2-dimethyldiselane, acmc-209ogy, methyl diselenide, 8ci | |
(methyldiselanyl)methane |
Specifications
7101-31-7 | |
155°C to 157°C | |
Unpleasant | |
1.6388 | |
1 g | |
Air and moisture sensitive | |
Reacts with water. | |
C[Se][Se]C | |
188.012 | |
187.99 | |
Liquid |
1.987 g/mL | |
60°C (140°F) | |
C2H6Se2 | |
MFCD00008254 | |
UN2929 | |
dimethyl diselenide, dimethyldiselenide, methyl diselenide, diselenide, dimethyl, ch3se 2, methyldiselanyl methane, diselenobismethane, 1,2-dimethyldiselane, acmc-209ogy, methyl diselenide, 8ci | |
VLXBWPOEOIIREY-UHFFFAOYSA-N | |
(methyldiselanyl)methane | |
23496 | |
99% | |
Dimethyl diselenide |
Safety and Handling
GHS H Statement
H301-H331-H226-H373
Toxic if swallowed.
Toxic if inhaled.
Flammable liquid and vapor.
May cause damage to organs through prolonged or repeated exposure.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.
EINECSNumber : 230-410-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only