missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl itaconate, 97%, stabilized, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 149160025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Dimethyl itaconate | |
| 1.2400g/mL | |
| 100°C | |
| 96% min. (GC) | |
| C7H10O4 | |
| MFCD00008446 | |
| 02,762 | |
| dimethyl itaconate, dimethyl 2-methylenesuccinate, dimethylitaconate, itaconic acid dimethyl ester, itaconic acid, dimethyl ester, butanedioic acid, methylene-, dimethyl ester, 1,4-dimethyl 2-methylidenebutanedioate, dimethyl methylenesuccinate, unii-11jib0yi93, succinic acid, methylene-, dimethyl ester | |
| ZWWQRMFIZFPUAA-UHFFFAOYSA-N | |
| dimethyl 2-methylidenebutanedioate | |
| 69240 | |
| 97% |
| 617-52-7 | |
| 208.0°C | |
| Authentic | |
| Glass bottle | |
| CH3O2CCH2C(=CH2)CO2CH3 | |
| 2.5kg | |
| 1.24 | |
| Solubility in water: slightly soluble | |
| COC(=O)CC(=C)C(=O)OC | |
| 158.15 | |
| 158.15 |
Safety and Handling
EINECSNumber : 210-519-6
RUO â Research Use Only