missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethyl malonate, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A110070E

 View more versions of this product

Catalog No. AAA110070E


Explore available promotions

Description

Description

Dimethyl malonate is a reagent used in organic synthesis. It acts as a precursor for the synthesis of mono-substituted and di-substituted acetic acid, barbiturates, vitamin B1 and vitamin B6. It is used in the pharmaceutical industry to prepare pharmaceuticals like chloroquine and butazolidin. It is involved in the synthesis of diastereomeric pure thienylpyridines by reacting with 1,3-diphenylprop-2-enyl acetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl malonate is a reagent used in organic synthesis. It acts as a precursor for the synthesis of mono-substituted and di-substituted acetic acid, barbiturates, vitamin B1 and vitamin B6. It is used in the pharmaceutical industry to prepare pharmaceuticals like chloroquine and butazolidin. It is involved in the synthesis of diastereomeric pure thienylpyridines by reacting with 1,3-diphenylprop-2-enyl acetate.

Solubility
Miscible with alcohol, ether and oils. Slightly miscible with water.

Notes
Incompatible with acids, bases, oxidizing agents and reducing agents.
Specifications

Specifications

Dimethyl malonate
-62°C
180°C to 181°C
C5H8O4
MFCD00008460
774261
dimethyl malonate, methyl malonate, propanedioic acid, dimethyl ester, malonic acid dimethyl ester, 1,3-dimethyl propanedioate, dimethyl 1,3-propanedioate, malonic acid, dimethyl ester, unii-em8y79998c, ccris 8981, propanedioic acid dimethyl ester
BEPAFCGSDWSTEL-UHFFFAOYSA-N
dimethyl propanedioate
7943
≥98%
108-59-8
1.156
90°C (194°F)
1.413
2500 g
14,6096
Miscible with alcohol,ether and oils. Slightly miscible with water.
COC(=O)CC(=O)OC
132.115
132.12

Chemical Identifiers

108-59-8
132.115
BEPAFCGSDWSTEL-UHFFFAOYSA-N
7943
COC(=O)CC(=O)OC
C5H8O4
MFCD00008460
dimethyl malonate, methyl malonate, propanedioic acid, dimethyl ester, malonic acid dimethyl ester, 1,3-dimethyl propanedioate, dimethyl 1,3-propanedioate, malonic acid, dimethyl ester, unii-em8y79998c, ccris 8981, propanedioic acid dimethyl ester
dimethyl propanedioate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P233-P240-P241-P242-P243-P264b-P280-P305+P351+P338-P374-P501c

H227-H320

EINECSNumber : 203-597-8

RTECSNumber : OO0950000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only