missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethyl malonate, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 220700010

 View more versions of this product

Catalog No. AC220700010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Dimethyl malonate
108-59-8
Colorless
180.0°C to 181.0°C
Authentic
Glass bottle
1.4120 to 1.4140
MFCD00008460
02,572
1.156
dimethyl malonate, methyl malonate, propanedioic acid, dimethyl ester, malonic acid dimethyl ester, 1,3-dimethyl propanedioate, dimethyl 1,3-propanedioate, malonic acid, dimethyl ester, unii-em8y79998c, ccris 8981, propanedioic acid dimethyl ester
BEPAFCGSDWSTEL-UHFFFAOYSA-N
dimethyl propanedioate
7943
99+%
99+%
-62.0°C
1.1560g/mL
90°C
99% min. (GC)
C5H8O4
CH3O2CCH2CO2CH3
1 L
08,193; 13,34
15,6168
Solubility in water: negligible. Other solubilities: soluble in alcohol and ether
COC(=O)CC(=O)OC
132.12
132.12
Liquid

Chemical Identifiers

108-59-8
132.12
BEPAFCGSDWSTEL-UHFFFAOYSA-N
7943
COC(=O)CC(=O)OC
C5H8O4
MFCD00008460
dimethyl malonate, methyl malonate, propanedioic acid, dimethyl ester, malonic acid dimethyl ester, 1,3-dimethyl propanedioate, dimethyl 1,3-propanedioate, malonic acid, dimethyl ester, unii-em8y79998c, ccris 8981, propanedioic acid dimethyl ester
dimethyl propanedioate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 203-597-8

RTECSNumber : OO0950000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only