missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dimethyl malonate, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 220702500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Dimethyl malonate | |
108-59-8 | |
Colorless | |
180.0°C to 181.0°C | |
Authentic | |
Glass bottle | |
1.4120 to 1.4140 | |
MFCD00008460 | |
02,572 | |
1.156 | |
dimethyl malonate, methyl malonate, propanedioic acid, dimethyl ester, malonic acid dimethyl ester, 1,3-dimethyl propanedioate, dimethyl 1,3-propanedioate, malonic acid, dimethyl ester, unii-em8y79998c, ccris 8981, propanedioic acid dimethyl ester | |
BEPAFCGSDWSTEL-UHFFFAOYSA-N | |
dimethyl propanedioate | |
7943 | |
99+% |
99+% | |
-62.0°C | |
1.1560g/mL | |
90°C | |
99% min. (GC) | |
C5H8O4 | |
CH3O2CCH2CO2CH3 | |
250 mL | |
08,193; 13,34 | |
15,6168 | |
Solubility in water: negligible. Other solubilities: soluble in alcohol and ether | |
COC(=O)CC(=O)OC | |
132.12 | |
132.12 | |
Liquid |
Chemical Identifiers
108-59-8 | |
132.12 | |
BEPAFCGSDWSTEL-UHFFFAOYSA-N | |
7943 | |
COC(=O)CC(=O)OC |
C5H8O4 | |
MFCD00008460 | |
dimethyl malonate, methyl malonate, propanedioic acid, dimethyl ester, malonic acid dimethyl ester, 1,3-dimethyl propanedioate, dimethyl 1,3-propanedioate, malonic acid, dimethyl ester, unii-em8y79998c, ccris 8981, propanedioic acid dimethyl ester | |
dimethyl propanedioate |
Safety and Handling
EINECSNumber : 203-597-8
RTECSNumber : OO0950000
TSCA : TSCA
RUO – Research Use Only