Learn More
Dimethyl succinate, 98%, Thermo Scientific Chemicals
$71.82 - $439.08
Chemical Identifiers
CAS | 106-65-0 |
---|---|
Molecular Formula | C6H10O4 |
Molecular Weight (g/mol) | 146.142 |
MDL Number | MFCD00008466 |
InChI Key | MUXOBHXGJLMRAB-UHFFFAOYSA-N |
Synonym | dimethyl succinate, methyl succinate, dimethylsuccinate, butanedioic acid, dimethyl ester, succinic acid, dimethyl ester, dbe-4 dibasic ester, methyl butanedioate, succinic acid dimethyl ester, butanedioic acid, 1,4-dimethyl ester, fema no. 2396 |
PubChem CID | 7820 |
IUPAC Name | dimethyl butanedioate |
SMILES | COC(=O)CCC(=O)OC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1256522
|
Thermo Scientific Chemicals
A1256522 |
100 g |
Each for $71.82
|
|
|||||
AAA1256530
|
Thermo Scientific Chemicals
A1256530 |
250 g |
Each for $134.75
|
|
|||||
AAA125650B
|
Thermo Scientific Chemicals
A125650B |
1000 g |
Each for $439.08
|
|
|||||
Description
Dimethyl succinate is used as a flavoring agent. It have wide range of industrial applications such as Functional fluids (open systems), Intermediates, Paint additives and coating additives, Pigments Solvents, Viscosity adjustors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDimethyl succinate is used as a flavoring agent. It have wide range of industrial applications such as Functional fluids (open systems), Intermediates, Paint additives and coating additives, Pigments Solvents, Viscosity adjustors.
Solubility
Soluble in water. (122,9 g/L at 20°C). soluble in acetone, ethanol, very soluble in ethyl ether.
Notes
Store at room temperature. Ensure good ventilation.
Chemical Identifiers
106-65-0 | |
146.142 | |
MUXOBHXGJLMRAB-UHFFFAOYSA-N | |
7820 | |
COC(=O)CCC(=O)OC |
C6H10O4 | |
MFCD00008466 | |
dimethyl succinate, methyl succinate, dimethylsuccinate, butanedioic acid, dimethyl ester, succinic acid, dimethyl ester, dbe-4 dibasic ester, methyl butanedioate, succinic acid dimethyl ester, butanedioic acid, 1,4-dimethyl ester, fema no. 2396 | |
dimethyl butanedioate |
Specifications
106-65-0 | |
1.119 | |
85°C (185°F) | |
1.419 | |
100 g | |
14,8869 | |
Soluble in water. (122,9g/L at 20°C). soluble in acetone,ethanol,very soluble in ethyl ether. | |
COC(=O)CCC(=O)OC | |
146.142 | |
146.14 | |
Dimethyl succinate |
18°C to 19°C | |
196°C | |
C6H10O4 | |
MFCD00008466 | |
956776 | |
dimethyl succinate, methyl succinate, dimethylsuccinate, butanedioic acid, dimethyl ester, succinic acid, dimethyl ester, dbe-4 dibasic ester, methyl butanedioate, succinic acid dimethyl ester, butanedioic acid, 1,4-dimethyl ester, fema no. 2396 | |
MUXOBHXGJLMRAB-UHFFFAOYSA-N | |
dimethyl butanedioate | |
7820 | |
≥98% |
Safety and Handling
GHS H Statement
H319-H227
Causes serious eye irritation.
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 203-419-9
RTECSNumber : WM7675000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only