Learn More
Dimethyl sulfide, 99+%, Thermo Scientific Chemicals
$71.77 - $134.40
Chemical Identifiers
CAS | 75-18-3 |
---|---|
Molecular Formula | C2H6S |
Molecular Weight (g/mol) | 62.13 |
MDL Number | MFCD00008562 |
InChI Key | QMMFVYPAHWMCMS-UHFFFAOYSA-N |
Synonym | dimethyl sulfide, methyl sulfide, methane, thiobis, dimethylsulfide, dimethylsulphide, methyl thioether, 2-thiapropane, dimethyl sulphide, dimethyl thioether, methylthiomethane |
PubChem CID | 1068 |
ChEBI | CHEBI:17437 |
SMILES | CSC |
Description
Dimethyl sulfide is used as a displaceable ligand in chloro(dimethyl sulfide)gold(I) and other coordination compounds, as a reducing agent in ozonolysis reactions as well as a food flavoring component. Further, it is utilized for the preparation of dimethyl sulfoxide, which is a common and important industrial solvent. It finds application in the petroleum industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
75-18-3 | |
62.13 | |
QMMFVYPAHWMCMS-UHFFFAOYSA-N | |
1068 | |
CSC |
C2H6S | |
MFCD00008562 | |
dimethyl sulfide, methyl sulfide, methane, thiobis, dimethylsulfide, dimethylsulphide, methyl thioether, 2-thiapropane, dimethyl sulphide, dimethyl thioether, methylthiomethane | |
CHEBI:17437 |
Specifications
75-18-3 | |
0.845 g/mL | |
-48°C (-54°F) | |
C2H6S | |
MFCD00008562 | |
UN1164 | |
dimethyl sulfide, methyl sulfide, methane, thiobis, dimethylsulfide, dimethylsulphide, methyl thioether, 2-thiapropane, dimethyl sulphide, dimethyl thioether, methylthiomethane | |
QMMFVYPAHWMCMS-UHFFFAOYSA-N | |
(methylsulfanyl)methane | |
1068 | |
62.14 | |
Liquid |
-83°C | |
38°C | |
Unpleasant | |
1.4351 | |
250 mL | |
14,6123 | |
Miscible with alcohols,ethers,esters,ketones,aliphatic and aromatic hydrocarbons. Slightly miscible with | |
CSC | |
62.13 | |
CHEBI:17437 | |
≥99% | |
Dimethyl sulfide |
Safety and Handling
GHS H Statement
H225-H318-H302-H335-H315
Highly flammable liquid and vapor.
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
H225-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: DIMETHYL SULFIDE
EINECSNumber : 200-846-2
RTECSNumber : PV5075000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only