missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dimethyl Sulfoxide, ≥99% (Molecular Biology/ACS Certified), MP Biomedicals
Supplier: MP Biomedicals Inc 0219141880
Specifications
Dimethyl Sulfoxide | |
100 mL | |
MFCD00002089 | |
IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
methanesulfinylmethane | |
679 | |
78.13 |
67-68-5 | |
C2H6OS | |
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide | |
CS(C)=O | |
78.13 | |
CHEBI:28262 | |
Certified ACS Reagent |
Chemical Identifiers
67-68-5 | |
78.13 | |
IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
679 | |
CS(C)=O |
C2H6OS | |
MFCD00002089 | |
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide | |
CHEBI:28262 |