Learn More
Dimethyl Sulfoxide (Certified ACS), Fisher Chemical™
Note: Sold for nonmedical use only. Not to be used or sold as a drug for humans or animals
Supplier: Fisher Chemical BPD1284
Specifications
| Dimethyl Sulfoxide | |
| 67-68-5 | |
| Clear, Colorless Liquid | |
| 1.100g/cm³ | |
| 4 L | |
| Pass Test (USP, EP/BP) | |
| Liquid | |
| 99.9 % (CH3)2SO Minimum | |
| C2H6OS | |
| MFCD00002089 | |
| 1.98 mPa/s at 25°C | |
| dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide | |
| IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
| methanesulfinylmethane | |
| 679 | |
| 78.13 | |
| Certified ACS Reagent |
| 18.4°C | |
| Pass Test (USP) | |
| >190°C | |
| 189°C | |
| 87°C (188.6°F) | |
| HDPE Bottle | |
| 0.01% max. | |
| 0.001mEq/g max. | |
| (CH3)2SO | |
| 0.55mbar at 20°C | |
| 0.1% max. | |
| Soluble starch and sulfate: Pass Test (USP), 15 ppm max. (EP/BP) | |
| CS(C)=O | |
| 78.13 | |
| CHEBI:28262 | |
| ≥99.9% | |
| Dextrin: Pass Test (USP, EP/BP) |
Chemical Identifiers
| 67-68-5 | |
| 78.13 | |
| IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
| 679 | |
| methanesulfinylmethane |
| C2H6OS | |
| MFCD00002089 | |
| dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide | |
| CHEBI:28262 | |
| CS(C)=O |
Safety and Handling
WARNING!
Emergency Overview
Combustible liquid. DMSO readily penetrates skin and may carry other dissolved chemicals into the body. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur.
NFPA
Health:2
Flammability:2
Instability:0