missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethyl Sulfoxide, MilliporeSigma™

ACS Reagent Grade

$66.31 - $135.45

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
SMILES CS(C)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
317275100ML
View Documents
MilliporeSigma
317275100ML
100 mL Glass Bottle
Each for $66.31
 
317275500ML
View Documents
MilliporeSigma
317275500ML
500 mL Glass Bottle
Each for $135.45
 
Description

Description

Specifications

Chemical Identifiers

67-68-5
78.13
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
679
CS(C)=O
C2H6OS
MFCD00002089
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
CHEBI:28262

Specifications

18.5°C
Colorless
189°C
87°C (189°F)
Glass Bottle
C2H6OS
0.08 kPa (0.6mm Hg) (at 20°C)
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
methanesulfinylmethane
679
78.13
ACS Reagent
67-68-5
1.10g/cm3 at 20°C
100 mL
300°to302°C (572°to576°F)
Liquid
MFCD00002089
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
CS(C)=O
78.13
CHEBI:28262
≥99%
Dimethyl Sulfoxide
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store in a segregated and approved area. Keep container in a cool, well-ventilated area. Keep container tightly closed and sealed until ready for use. Avoid all possible sources of ignition (spark or flame).

SDS
missing translation for 'documents'

missing translation for 'documents'