missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethyl trisulfide, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H3291314

 View more versions of this product

Catalog No. AAH3291314


Explore available promotions

Description

Description

Dimethyl trisulfide is reported as a major aroma component in cooked Brassicaceous vegetables. Trap baits containing dimethyl trisulfide have been used to capture Calliphora loewi and other blowflies. Dimethyl trisulfide along with dimethyl sulfide and dimethyl disulfide have been confirmed as volatile compounds given off by the fly-attracting plant known as dead-horse arum (Helicodiceros muscivorus). These flies are attracted to the odor of fetid meat and help pollinate this plant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl trisulfide is reported as a major aroma component in cooked Brassicaceous vegetables. Trap baits containing dimethyl trisulfide have been used to capture Calliphora loewi and other blowflies. Dimethyl trisulfide along with dimethyl sulfide and dimethyl disulfide have been confirmed as volatile compounds given off by the fly-attracting plant known as dead-horse arum (Helicodiceros muscivorus). These flies are attracted to the odor of fetid meat and help pollinate this plant.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at 4°C. Store in an air-tight and light-resistant container.
Specifications

Specifications

Dimethyl trisulfide
1.203
52°C (126°F)
C2H6S3
CH3SSSCH3
25 g
dimethyl trisulfide, trisulfide, dimethyl, methyl trisulfide, 2,3,4-trithiapentane, dimethyl trisulphide, dimethyltrisulfide, methyltrisulfanyl methane, dimethyltrisulfane, unii-3e691t3nl1, dmts
YWHLKYXPLRWGSE-UHFFFAOYSA-N
(methyltrisulfanyl)methane
19310
126.26
3658-80-8
165°C to 175°C
Stench
1.6
MFCD00039808
UN1993
Insoluble in water.
CSSSC
126.25
CHEBI:4614
98%

Chemical Identifiers

3658-80-8
126.25
YWHLKYXPLRWGSE-UHFFFAOYSA-N
19310
(methyltrisulfanyl)methane
C2H6S3
MFCD00039808
dimethyl trisulfide, trisulfide, dimethyl, methyl trisulfide, 2,3,4-trithiapentane, dimethyl trisulphide, dimethyltrisulfide, methyltrisulfanyl methane, dimethyltrisulfane, unii-3e691t3nl1, dmts
CHEBI:4614
CSSSC
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c

H226-H302-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 222-910-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only