missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dimethyl Trisulfide 98.0+%, TCI America™
$64.02 - $188.59
Chemical Identifiers
CAS | 3658-80-8 |
---|---|
Molecular Formula | C2H6S3 |
Molecular Weight (g/mol) | 126.25 |
MDL Number | MFCD00039808 |
InChI Key | YWHLKYXPLRWGSE-UHFFFAOYSA-N |
Synonym | dimethyl trisulfide, trisulfide, dimethyl, methyl trisulfide, 2,3,4-trithiapentane, dimethyl trisulphide, dimethyltrisulfide, methyltrisulfanyl methane, dimethyltrisulfane, unii-3e691t3nl1, dmts |
PubChem CID | 19310 |
ChEBI | CHEBI:4614 |
IUPAC Name | (methyltrisulfanyl)methane |
SMILES | CSSSC |
Chemical Identifiers
3658-80-8 | |
126.25 | |
YWHLKYXPLRWGSE-UHFFFAOYSA-N | |
19310 | |
(methyltrisulfanyl)methane |
C2H6S3 | |
MFCD00039808 | |
dimethyl trisulfide, trisulfide, dimethyl, methyl trisulfide, 2,3,4-trithiapentane, dimethyl trisulphide, dimethyltrisulfide, methyltrisulfanyl methane, dimethyltrisulfane, unii-3e691t3nl1, dmts | |
CHEBI:4614 | |
CSSSC |
Specifications
3658-80-8 | |
Yellow | |
C2H6S3 | |
5 g | |
dimethyl trisulfide, trisulfide, dimethyl, methyl trisulfide, 2,3,4-trithiapentane, dimethyl trisulphide, dimethyltrisulfide, methyltrisulfanyl methane, dimethyltrisulfane, unii-3e691t3nl1, dmts | |
CSSSC | |
126.25 | |
CHEBI:4614 | |
≥98.0% (GC) | |
Dimethyl Trisulfide |
-68°C | |
170°C | |
MFCD00039808 | |
1993 | |
YWHLKYXPLRWGSE-UHFFFAOYSA-N | |
(methyltrisulfanyl)methane | |
19310 | |
126.25 | |
Liquid |
Safety and Handling
TSCA : Yes