Learn More
Dimethylaluminium chloride, 0.9M solution in heptane, AcroSeal™, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 377791000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidSpecifications
Dimethylaluminium chloride | |
1184-58-3,142-82-5 | |
0.7110g/mL | |
AcroSeal™ Glass bottle | |
MFCD00000458 | |
0.711 | |
aluminum, chlorodimethyl, dimethylaluminum chloride, dimethylaluminium chloride, chlorodimethylaluminum, dimethylaluminum chloride solution, 1.0 m in hexanes, chloro dimethyl alumane, dimethylaluminum chloride solution | |
JGHYBJVUQGTEEB-UHFFFAOYSA-M | |
chloro(dimethyl)alumane | |
79147 | |
Solution |
0.9M Solution in Heptane | |
Colorless to Gray | |
−4°C | |
C2H6AlCl | |
100 mL | |
15, 3249 | |
Solubility in water: vigorous reaction. Other solubilities: miscible with saturated aliphatic and aromatic,hydrocarbons | |
C[Al](C)Cl | |
92.5 | |
92.5 |
Chemical Identifiers
1184-58-3 | |
JGHYBJVUQGTEEB-UHFFFAOYSA-M | |
79147 | |
C[Al](C)Cl |
MFCD00000458 | |
aluminum, chlorodimethyl, dimethylaluminum chloride, dimethylaluminium chloride, chlorodimethylaluminum, dimethylaluminum chloride solution, 1.0 m in hexanes, chloro dimethyl alumane, dimethylaluminum chloride solution | |
chloro(dimethyl)alumane |
Safety and Handling
GHS H Statement
May cause drowsiness or dizziness.
Toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
Causes severe skin burns and eye damage.
In contact with water releases flamm
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap
GHS Signal Word: Danger
EINECSNumber : 214-668-8
RUO – Research Use Only