missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethylamine hydrochloride, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 155270050

 View more versions of this product

Catalog No. AC155270050

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Dimethylamine hydrochloride
506-59-2
White
Authentic
Glass bottle
(CH3)2NH·HCl
5 g
15, 3250
Solubility in water: 3000g/L (20°C). Other solubilities: soluble in alcohol,chloroform,practically insoluble in ether
CNC.Cl
81.55
81.55
0.005% max. (in water)
99%
160.0°C
5 to 6 (5% aq. soln.)
98.5% min. (on dry substance) (Argentometry)
C2H7N·HCl
MFCD00012477
04, 39
dimethylamine hydrochloride, n-methylmethanamine hydrochloride, dimethylammonium chloride, methanamine, n-methyl-, hydrochloride, dimethylamine hcl, dimethylaminehydrochloride, unii-7m4cwb6aok, dimethylamine, hydrochloride, hydrochloric acid dimethylamine, n,n-dimethylamine hydrochloride
IQDGSYLLQPDQDV-UHFFFAOYSA-N
N-methylmethanamine;hydrochloride
10473
99%
Crystalline Powder or Crystals

Chemical Identifiers

506-59-2
IQDGSYLLQPDQDV-UHFFFAOYSA-N
10473
MFCD00012477
dimethylamine hydrochloride, n-methylmethanamine hydrochloride, dimethylammonium chloride, methanamine, n-methyl-, hydrochloride, dimethylamine hcl, dimethylaminehydrochloride, unii-7m4cwb6aok, dimethylamine, hydrochloride, hydrochloric acid dimethylamine, n,n-dimethylamine hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 208-046-5

RTECSNumber : IQ0220000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only