missing translation for 'onlineSavingsMsg'
Learn More

Dimethylcyanamide, 97%, Thermo Scientific Chemicals

$66.21 - $440.95

Chemical Identifiers

CAS 1467-79-4
Molecular Formula C3H6N2
Molecular Weight (g/mol) 70.10
MDL Number MFCD00001767
InChI Key OAGOUCJGXNLJNL-UHFFFAOYSA-N
Synonym dimethyl cyanamide, cyanamide, dimethyl, n-cyano-n-methylmethanamine, dimethylkyanamid, cyanodimethylamine, n-cyanodimethylamine, dimethylkyanamid czech, unii-6p25ipq8gn, ccris 5909, cyanamide, n,n-dimethyl
PubChem CID 15112
IUPAC Name dimethylcyanamide
SMILES CN(C)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC200210050
SDS View Documents
Thermo Scientific Chemicals
200210050
5 mL Glass bottle
Each for $66.21
Only null left
 
AC200211000
SDS View Documents
Thermo Scientific Chemicals
200211000
100 mL Glass bottle
Each for $440.95
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1467-79-4
70.10
OAGOUCJGXNLJNL-UHFFFAOYSA-N
15112
CN(C)C#N
C3H6N2
MFCD00001767
dimethyl cyanamide, cyanamide, dimethyl, n-cyano-n-methylmethanamine, dimethylkyanamid, cyanodimethylamine, n-cyanodimethylamine, dimethylkyanamid czech, unii-6p25ipq8gn, ccris 5909, cyanamide, n,n-dimethyl
dimethylcyanamide

Specifications

1467-79-4
Colorless to Yellow
161.0°C to 163.0°C
Authentic
Glass bottle
1.4090 to 1.4110
MFCD00001767
04, 74
dimethyl cyanamide, cyanamide, dimethyl, n-cyano-n-methylmethanamine, dimethylkyanamid, cyanodimethylamine, n-cyanodimethylamine, dimethylkyanamid czech, unii-6p25ipq8gn, ccris 5909, cyanamide, n,n-dimethyl
OAGOUCJGXNLJNL-UHFFFAOYSA-N
dimethylcyanamide
15112
97%
Dimethylcyanamide, 97%
-41.0°C
0.8670g/mL
58°C
97%
C3H6N2
(CH3)2NCN
5 mL
0.867
Solubility in water: miscible. Other solubilities: soluble in acetone,ethyl ether,ethnaol
CN(C)C#N
70.10
70.09
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Fatal in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Toxic if inhaled.
Flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable f

GHS Signal Word: Danger

EINECSNumber : 215-991-7

RTECSNumber : GS6475000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only