missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dimethyloxaloylglycine, 98%, Thermo Scientific Chemicals
$70.25 - $484.15
Chemical Identifiers
CAS | 89464-63-1 |
---|---|
Molecular Formula | C6H9NO5 |
Molecular Weight (g/mol) | 175.14 |
MDL Number | MFCD05865098 |
InChI Key | BNJOZDZCRHCODO-UHFFFAOYSA-N |
Synonym | dmog, dimethyloxalylglycine, dimethyloxaloylglycine, dimethyloxallyl glycine, methyl n-methoxy oxo acetyl glycinate, methyl 2-2-methoxy-2-oxoacetamido acetate, methyl 2-2-methoxy-2-oxoethyl amino-2-oxoacetate, glycine, n-methoxyoxoacetyl-, methyl ester, n-2-methoxy-2-oxoacetyl glycine methyl ester, glycine,n-2-methoxy-2-oxoacetyl-, methyl ester |
PubChem CID | 560326 |
IUPAC Name | methyl 2-(2-methoxy-2-oxoacetamido)acetate |
SMILES | COC(=O)CNC(=O)C(=O)OC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ67077MA
|
Thermo Scientific Chemicals
J67077MA |
10 mg |
Each for $70.25
|
|
|||||
AAJ67077MF
|
Thermo Scientific Chemicals
J67077MF |
50 mg |
Each for $279.62
|
|
|||||
AAJ67077MC
|
Thermo Scientific Chemicals
J67077MC |
100 mg |
Each for $484.15
|
|
|||||
Description
DMOG is a cell permeable prolyl-4-hydroxylase inhibitor that regulates hypoxia-inducible factor
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
89464-63-1 | |
175.14 | |
BNJOZDZCRHCODO-UHFFFAOYSA-N | |
560326 | |
COC(=O)CNC(=O)C(=O)OC |
C6H9NO5 | |
MFCD05865098 | |
dmog, dimethyloxalylglycine, dimethyloxaloylglycine, dimethyloxallyl glycine, methyl n-methoxy oxo acetyl glycinate, methyl 2-2-methoxy-2-oxoacetamido acetate, methyl 2-2-methoxy-2-oxoethyl amino-2-oxoacetate, glycine, n-methoxyoxoacetyl-, methyl ester, n-2-methoxy-2-oxoacetyl glycine methyl ester, glycine,n-2-methoxy-2-oxoacetyl-, methyl ester | |
methyl 2-(2-methoxy-2-oxoacetamido)acetate |
Specifications
89464-63-1 | |
C6H9NO5 | |
dmog, dimethyloxalylglycine, dimethyloxaloylglycine, dimethyloxallyl glycine, methyl n-methoxy oxo acetyl glycinate, methyl 2-2-methoxy-2-oxoacetamido acetate, methyl 2-2-methoxy-2-oxoethyl amino-2-oxoacetate, glycine, n-methoxyoxoacetyl-, methyl ester, n-2-methoxy-2-oxoacetyl glycine methyl ester, glycine,n-2-methoxy-2-oxoacetyl-, methyl ester | |
BNJOZDZCRHCODO-UHFFFAOYSA-N | |
methyl 2-(2-methoxy-2-oxoacetamido)acetate | |
560326 | |
98% | |
Solid |
White | |
MFCD05865098 | |
Soluble in water at 30mg/ml. | |
COC(=O)CNC(=O)C(=O)OC | |
175.14 | |
175.14 | |
10 mg | |
Dimethyloxaloylglycine |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only