Learn More
Dimethyltin dichloride, Thermo Scientific Chemicals
$57.73 - $519.10
Chemical Identifiers
CAS | 753-73-1 |
---|---|
Molecular Formula | C2H6Cl2Sn |
Molecular Weight (g/mol) | 219.68 |
MDL Number | MFCD00000517 |
InChI Key | PKKGKUDPKRTKLJ-UHFFFAOYSA-L |
Synonym | dimethyltin dichloride, dimethyldichlorotin, stannane, dichlorodimethyl, dichlorodimethyltin, dimethyltindichloride, dimethyldichlorostannane, dichloro dimethyl stannane, tin, dimethyl-, dichloride, cotin 210, dichlorid dimethylcinicity |
PubChem CID | 12955 |
IUPAC Name | dichloro(dimethyl)stannane |
SMILES | C[Sn](C)(Cl)Cl |
Description
Used as a pharmaceutical intermediate. It is employed as a coating, plastic and rubber auxilliary agent. Used as methyl Tin stabilizer intermediates, catalysts for esterification reaction, raw material for glass coating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
753-73-1 | |
219.68 | |
PKKGKUDPKRTKLJ-UHFFFAOYSA-L | |
12955 | |
C[Sn](C)(Cl)Cl |
C2H6Cl2Sn | |
MFCD00000517 | |
dimethyltin dichloride, dimethyldichlorotin, stannane, dichlorodimethyl, dichlorodimethyltin, dimethyltindichloride, dimethyldichlorostannane, dichloro dimethyl stannane, tin, dimethyl-, dichloride, cotin 210, dichlorid dimethylcinicity | |
dichloro(dimethyl)stannane |
Specifications
753-73-1 | |
185°C to 190°C | |
MFCD00000517 | |
UN3146 | |
Moisture sensitive | |
Slightly soluble in water (20g/L (20°C)). Solubility in other solvents (0.1g/ml in methanol). | |
C[Sn](C)(Cl)Cl | |
219.68 | |
219.69 | |
Dimethyltin dichloride |
103°C to 105°C | |
C2H6Cl2Sn | |
2 g | |
3660508 | |
dimethyltin dichloride, dimethyldichlorotin, stannane, dichlorodimethyl, dichlorodimethyltin, dimethyltindichloride, dimethyldichlorostannane, dichloro dimethyl stannane, tin, dimethyl-, dichloride, cotin 210, dichlorid dimethylcinicity | |
PKKGKUDPKRTKLJ-UHFFFAOYSA-L | |
dichloro(dimethyl)stannane | |
12955 | |
Crystalline |
Safety and Handling
GHS H Statement
H300-H310-H330-H314-H318
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P201-P202-P260-P264b-P270-P271-P281-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H301+H311-H314-H330-H360-H372
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 212-039-2
RTECSNumber : WH7245000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only