missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dinonyl phthalate, 96%, mixture of isomers
CAS: 84-76-4 | C26H42O4 | 418.62 g/mol
$258.69 - $258.69
Identifiants chimiques
| CAS | 84-76-4 |
|---|---|
| Molecular Formula | C26H42O4 |
| Molecular Weight (g/mol) | 418.62 |
| MDL Number | MFCD00036237 |
| InChI Key | DROMNWUQASBTFM-UHFFFAOYSA-N |
| Synonym | dinonyl phthalate, dinonylphthalate, nonyl phthalate, di-n-nonyl phthalate, bisoflex dnp, unimoll dn, bisoflex 91, phthalic acid, dinonyl ester, 1,2-benzenedicarboxylic acid, dinonyl ester, bisolflex 91 |
| PubChem CID | 6787 |
| SMILES | CCCCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCCCC |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|
AC408770250
|
Thermo Scientific Chemicals
408770250 |
25 g | Glass bottle |
chaque for $258.69
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
| 84-76-4 | |
| 418.62 | |
| DROMNWUQASBTFM-UHFFFAOYSA-N | |
| 6787 |
| C26H42O4 | |
| MFCD00036237 | |
| dinonyl phthalate, dinonylphthalate, nonyl phthalate, di-n-nonyl phthalate, bisoflex dnp, unimoll dn, bisoflex 91, phthalic acid, dinonyl ester, 1,2-benzenedicarboxylic acid, dinonyl ester, bisolflex 91 | |
| CCCCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCCCC |
Spécifications
| 84-76-4 | |
| 0.9700g/mL | |
| 216°C | |
| 95% min. (sum of isomers) (GC) | |
| C26H42O4 | |
| MFCD00036237 | |
| 0.97 | |
| Solubility in water: <1g/L (20°) | |
| CCCCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCCCC | |
| 418.62 | |
| 166 mPa.s (20°C) | |
| 96% | |
| Dinonyl phthalate |
| Yellow to Brown | |
| 279.0°C to 287.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4825 to 1.4845 | |
| 25 g | |
| dinonyl phthalate, dinonylphthalate, nonyl phthalate, di-n-nonyl phthalate, bisoflex dnp, unimoll dn, bisoflex 91, phthalic acid, dinonyl ester, 1,2-benzenedicarboxylic acid, dinonyl ester, bisolflex 91 | |
| DROMNWUQASBTFM-UHFFFAOYSA-N | |
| 1,2-dinonyl benzene-1,2-dicarboxylate | |
| 6787 | |
| 418.61 | |
| Oily Liquid |
Sécurité et manipulation
missing translation for 'einecsNumber' : 201-560-
RUO – Research Use Only