missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diosmin, Thermo Scientific Chemicals
$69.22 - $189.92
Chemical Identifiers
CAS | 520-27-4 |
---|---|
Molecular Formula | C28H32O15 |
Molecular Weight (g/mol) | 608.549 |
MDL Number | MFCD00009772 |
InChI Key | GZSOSUNBTXMUFQ-YFAPSIMESA-N |
Synonym | diosmin, barosmin, diosmine, venosmine, diosmil, flebosten, daflon, diosimin, diosminum, tovene |
PubChem CID | 5281613 |
ChEBI | CHEBI:4631 |
IUPAC Name | 5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one |
SMILES | CC1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3=CC(=C4C(=C3)OC(=CC4=O)C5=CC(=C(C=C5)OC)O)O)O)O)O)O)O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6207306
|
Thermo Scientific Chemicals
J6207306 |
5 g |
Each for $69.22
|
|
|||||
AAJ6207314
|
Thermo Scientific Chemicals
J6207314 |
25 g |
Each for $189.92
|
|
|||||
Description
Diosmin exhibits anti-proliferative and anti-inflammatory. It is used in the treatment of venous disease, i.e., chronic venous insufficiency (CVI) and hemorrhoidal disease (HD).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in DMSO (50 mg/ml), water (122 mg/ml at 25°C), and ethanol (<1mg/mL at 25°C).
Notes
Protect from heat. Incompatible with heat and oxidizing agents. Store at 4°C.
Chemical Identifiers
520-27-4 | |
608.549 | |
GZSOSUNBTXMUFQ-YFAPSIMESA-N | |
5281613 | |
5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one |
C28H32O15 | |
MFCD00009772 | |
diosmin, barosmin, diosmine, venosmine, diosmil, flebosten, daflon, diosimin, diosminum, tovene | |
CHEBI:4631 | |
CC1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3=CC(=C4C(=C3)OC(=CC4=O)C5=CC(=C(C=C5)OC)O)O)O)O)O)O)O)O |
Specifications
520-27-4 | |
Brown to Yellow | |
MFCD00009772 | |
14,3297 | |
Soluble in DMSO (50mg/ml),water (122mg/ml at 25°C),and ethanol (<1mg/ml at 25°C). | |
CC1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3=CC(=C4C(=C3)OC(=CC4=O)C5=CC(=C(C=C5)OC)O)O)O)O)O)O)O)O | |
608.549 | |
CHEBI:4631 | |
Powder |
274°C | |
C28H32O15 | |
5 g | |
diosmin, barosmin, diosmine, venosmine, diosmil, flebosten, daflon, diosimin, diosminum, tovene | |
GZSOSUNBTXMUFQ-YFAPSIMESA-N | |
5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one | |
5281613 | |
608.5 | |
Diosmin |
Safety and Handling
EINECSNumber : 208-289-7
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only