missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dipentene, tech., Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 045805AK

 View more versions of this product

Catalog No. AA45805AK


Explore available promotions

Description

Description

Dipentene is a monoterpene olefin having potential applications in polymer and fuel chemistry. It is also widely used as flavoring and fragrance agent. It is a promising green solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Dipentene
0.85 g/mL
43°C (109°F)
1.474
MFCD00062992
UN2052
14,5493
Soluble in ether (miscible) and ethanol (miscible). Insoluble in water.
CC(=C)C1CCC(C)=CC1
136.24
CHEBI:15384
Technical
138-86-3
170°C to 180°C
C10H16
CH3C6H8C(CH3)=CH2
250 mL
3587825
dipentene, limonene, cinene, cajeputene, dl-limonene, kautschin, p-mentha-1,8-diene, dipenten, polylimonene, eulimen
XMGQYMWWDOXHJM-UHFFFAOYNA-N
1-methyl-4-prop-1-en-2-ylcyclohexene
22311
136.24
Liquid

Chemical Identifiers

138-86-3
136.24
XMGQYMWWDOXHJM-UHFFFAOYNA-N
22311
CC(=C)C1CCC(C)=CC1
C10H16
MFCD00062992
dipentene, limonene, cinene, cajeputene, dl-limonene, kautschin, p-mentha-1,8-diene, dipenten, polylimonene, eulimen
CHEBI:15384
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H317
Flammable liquid and vapor.
Causes skin irritation.
May cause an allergic skin reaction.

P210-P233-P240-P241-P242-P243-P261-P264b-P272-P280-P303+P361+P353-P305+P351+P338-P333+P313-P363-P370+P378q-P501c

H226-H315-H317-H319

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: DIPENTENE

EINECSNumber : 205-341-0

RTECSNumber : OS8100000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only