missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl-2-pyridylphosphine, 97%

CAS: 37943-90-1 | C17H14NP | 263.28 g/mol

$244.58 - $244.58

Chemical Identifiers

CAS 37943-90-1
Molecular Formula C17H14NP
Molecular Weight (g/mol) 263.28
MDL Number MFCD00192108
InChI Key SVABQOITNJTVNJ-UHFFFAOYSA-N
Synonym diphenyl-2-pyridylphosphine, 2-diphenylphosphino pyridine, 2-pyridyldiphenylphosphine, diphenyl 2-pyridinyl phosphine, diphenyl 2-pyridyl phosphane, 2-diphenylphosphanyl-pyridine, 2-diphenylphosphanyl pyridine, diphenyl pyridin-2-yl phosphane, pph2 2-pyridyl, dpppy
PubChem CID 621893
IUPAC Name diphenyl(pyridin-2-yl)phosphane
SMILES C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=N3
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC427660050
SDS View Documents
Thermo Scientific Chemicals
427660050
5 g Glass bottle
Each for $244.58
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

37943-90-1
263.28
SVABQOITNJTVNJ-UHFFFAOYSA-N
621893
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=N3
C17H14NP
MFCD00192108
diphenyl-2-pyridylphosphine, 2-diphenylphosphino pyridine, 2-pyridyldiphenylphosphine, diphenyl 2-pyridinyl phosphine, diphenyl 2-pyridyl phosphane, 2-diphenylphosphanyl-pyridine, 2-diphenylphosphanyl pyridine, diphenyl pyridin-2-yl phosphane, pph2 2-pyridyl, dpppy
diphenyl(pyridin-2-yl)phosphane

Specifications

37943-90-1
Tan to White
Authentic
Glass bottle
MFCD00192108
diphenyl-2-pyridylphosphine, 2-diphenylphosphino pyridine, 2-pyridyldiphenylphosphine, diphenyl 2-pyridinyl phosphine, diphenyl 2-pyridyl phosphane, 2-diphenylphosphanyl-pyridine, 2-diphenylphosphanyl pyridine, diphenyl pyridin-2-yl phosphane, pph2 2-pyridyl, dpppy
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=N3
263.28
263.28
Crystalline Powder
82.0°C to 84.0°C
74.85 to 80.27% C; 5.17 to 5.55% H; 5.1 to 5.6% N
97%
C17H14NP
5 g
SVABQOITNJTVNJ-UHFFFAOYSA-N
diphenyl(pyridin-2-yl)phosphane
621893
97%
Diphenyl-2-pyridylphosphine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only