Learn More
Diphenyl disulfide, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1258630
Description
Diphenyl disulfide is one of the most popular organic disulfides used in organic synthesis. It is used as a reagent for the α-phenylsulfenylation of carbonyl compounds. It participates in hydrothiolation of alkynes via amine-mediated single electron transfer mechanism. It is the hydrolysis product of dyfonate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiphenyl disulfide is one of the most popular organic disulfides used in organic synthesis. It is used as a reagent for the α-phenylsulfenylation of carbonyl compounds. It participates in hydrothiolation of alkynes via amine-mediated single electron transfer mechanism. It is the hydrolysis product of dyfonate.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Diphenyl disulfide | |
58°C to 62°C | |
Unpleasant | |
MFCD00003065 | |
639794 | |
Insoluble in water. | |
C1=CC=C(C=C1)SSC2=CC=CC=C2 | |
218.332 | |
218.34 |
882-33-7 | |
310°C | |
C12H10S2 | |
250 g | |
diphenyl disulfide, phenyl disulfide, disulfide, diphenyl, diphenyldisulfide, 1,2-diphenyldisulfane, phenyldisulfanyl benzene, diphenyl disulphide, biphenyl disulfide, disulfide diphenyl, phenyldithiobenzene | |
GUUVPOWQJOLRAS-UHFFFAOYSA-N | |
(phenyldisulfanyl)benzene | |
13436 | |
98% |
Chemical Identifiers
882-33-7 | |
218.332 | |
GUUVPOWQJOLRAS-UHFFFAOYSA-N | |
13436 | |
C1=CC=C(C=C1)SSC2=CC=CC=C2 |
C12H10S2 | |
MFCD00003065 | |
diphenyl disulfide, phenyl disulfide, disulfide, diphenyl, diphenyldisulfide, 1,2-diphenyldisulfane, phenyldisulfanyl benzene, diphenyl disulphide, biphenyl disulfide, disulfide diphenyl, phenyldithiobenzene | |
(phenyldisulfanyl)benzene |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 212-926-4
RTECSNumber : SS6825000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only