missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diphenyl Oxalate 98.0+%, TCI America™
Supplier: TCI America O0111100G
Specifications
Diphenyl Oxalate | |
136°C | |
MFCD00059682 | |
Oxalic Acid Diphenyl Ester | |
C1=CC=C(C=C1)OC(=O)C(=O)OC2=CC=CC=C2 | |
242.23 | |
242.23 | |
Crystalline Powder |
3155-16-6 | |
C14H10O4 | |
100 g | |
ULOZDEVJRTYKFE-UHFFFAOYSA-N | |
diphenyl oxalate | |
18475 | |
≥98.0% (GC) |
Chemical Identifiers
3155-16-6 | |
242.23 | |
ULOZDEVJRTYKFE-UHFFFAOYSA-N | |
18475 | |
C1=CC=C(C=C1)OC(=O)C(=O)OC2=CC=CC=C2 |
C14H10O4 | |
MFCD00059682 | |
Oxalic Acid Diphenyl Ester | |
diphenyl oxalate |
Safety and Handling
RTECSNumber : RO2880000
TSCA : No