missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diphenyl phosphate, 99%, Thermo Scientific Chemicals

$79.27 - $79.27

Chemical Identifiers

CAS 838-85-7
Molecular Formula C12H11O4P
Molecular Weight (g/mol) 250.19
MDL Number MFCD00003033
InChI Key ASMQGLCHMVWBQR-UHFFFAOYSA-N
Synonym diphenyl phosphate, phosphoric acid, diphenyl ester, phenyl hydrogen phosphate, phenyl phosphate pho 2 ho po, diphenoxyphosphinic acid, phosphoric acid diphenyl ester, diphenylphosphoric acid, phosphoric acid diphenyl, dsstox_cid_28182
PubChem CID 13282
IUPAC Name diphenyl hydrogen phosphate
SMILES OP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC225690050
SDS View Documents
Thermo Scientific Chemicals
225690050
5 g Glass bottle
Each for $79.27
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

838-85-7
250.19
ASMQGLCHMVWBQR-UHFFFAOYSA-N
13282
OP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1
C12H11O4P
MFCD00003033
diphenyl phosphate, phosphoric acid, diphenyl ester, phenyl hydrogen phosphate, phenyl phosphate pho 2 ho po, diphenoxyphosphinic acid, phosphoric acid diphenyl ester, diphenylphosphoric acid, phosphoric acid diphenyl, dsstox_cid_28182
diphenyl hydrogen phosphate

Specifications

838-85-7
White
99%
C12H11O4P
MFCD00003033
06, 178
ASMQGLCHMVWBQR-UHFFFAOYSA-N
diphenyl hydrogen phosphate
13282
99%
Diphenyl phosphate, 99%
63°C to 70°C
Authentic
Glass bottle
(C6H5O)2P(O)OH
5 g
diphenyl phosphate, phosphoric acid, diphenyl ester, phenyl hydrogen phosphate, phenyl phosphate pho 2 ho po, diphenoxyphosphinic acid, phosphoric acid diphenyl ester, diphenylphosphoric acid, phosphoric acid diphenyl, dsstox_cid_28182
OP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1
250.19
250.19
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 212-657-2

RTECSNumber : TC5470000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only