missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl Phosphite (contains ca. 5% Phenol) 75.0+%, TCI America™

$29.83 - $115.57

Chemical Identifiers

CAS 4712-55-4
Molecular Formula C12H10O3P+
Molecular Weight (g/mol) 233.183
MDL Number MFCD00044497
InChI Key CDXVUROVRIFQMV-UHFFFAOYSA-N
Synonym diphenyl phosphite, diphenyl phosphonate, phosphonic acid, diphenyl ester, diphenoxyphosphine oxide, diphenyl hydrogen phosphite, phosphorous acid, diphenyl ester, unii-5144js6xum, phosphonic acid diphenyl ester, phenyl phosphonate, pho 2hpo, phenyl phosphonate pho 2hpo
PubChem CID 6327546
IUPAC Name oxo(diphenoxy)phosphanium
SMILES C1=CC=C(C=C1)O[P+](=O)OC2=CC=CC=C2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D090725G
View Documents
TCI America
D090725G
25 g
Each for $29.83
Only null left
 
D0907100G
View Documents
TCI America
D0907100G
100 g
Each for $52.19
Only null left
 
D0907500G
View Documents
TCI America
D0907500G
500 g
Each for $115.57
Only null left
 
Description

Description

Specifications

Chemical Identifiers

4712-55-4
233.183
CDXVUROVRIFQMV-UHFFFAOYSA-N
6327546
C1=CC=C(C=C1)O[P+](=O)OC2=CC=CC=C2
C12H10O3P+
MFCD00044497
diphenyl phosphite, diphenyl phosphonate, phosphonic acid, diphenyl ester, diphenoxyphosphine oxide, diphenyl hydrogen phosphite, phosphorous acid, diphenyl ester, unii-5144js6xum, phosphonic acid diphenyl ester, phenyl phosphonate, pho 2hpo, phenyl phosphonate pho 2hpo
oxo(diphenoxy)phosphanium

Specifications

4712-55-4
Yellow
C12H10O3P+
25 g
diphenyl phosphite, diphenyl phosphonate, phosphonic acid, diphenyl ester, diphenoxyphosphine oxide, diphenyl hydrogen phosphite, phosphorous acid, diphenyl ester, unii-5144js6xum, phosphonic acid diphenyl ester, phenyl phosphonate, pho 2hpo, phenyl phosphonate pho 2hpo
C1=CC=C(C=C1)O[P+](=O)OC2=CC=CC=C2
233.183
234.19
Liquid
12°C
148°C
MFCD00044497
1760
CDXVUROVRIFQMV-UHFFFAOYSA-N
oxo(diphenoxy)phosphanium
6327546
≥75.0% (HPLC)
Diphenyl Phosphite (contains ca. 5% Phenol)
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2602

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'