Learn More
Diphenylacetic acid, 99%, Thermo Scientific Chemicals
$96.71 - $385.47
Chemical Identifiers
CAS | 117-34-0 |
---|---|
Molecular Formula | C14H12O2 |
Molecular Weight (g/mol) | 212.248 |
MDL Number | MFCD00004251 |
InChI Key | PYHXGXCGESYPCW-UHFFFAOYSA-N |
Synonym | diphenylacetic acid, diphenylethanoic acid, acetic acid, diphenyl, benzeneacetic acid, .alpha.-phenyl, 1,1-diphenylacetic acid, unii-658ncz0nko, diphenyl-acetic acid, alpha-toluic acid, alpha-phenyl, 658ncz0nko, benzeneacetic acid, alpha-phenyl |
PubChem CID | 8333 |
ChEBI | CHEBI:41967 |
IUPAC Name | 2,2-diphenylacetic acid |
SMILES | C1=CC=C(C=C1)C(C2=CC=CC=C2)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1148922
|
Thermo Scientific Chemicals
A1148922 |
100 g |
Each for $96.71
|
|
|||||
AAA1148936
|
Thermo Scientific Chemicals
A1148936 |
500 g |
Each for $385.47
|
|
|||||
Description
Kinetic resolution of racemic 1-heteroarylalkanols by asymmetric esterification using diphenylacetic acid with pivalic anhydride and a chiral acyl-transfer catalyst is described. Molecular crystal of acridine and diphenylacetic acid and its absolute asymmetric photodecarboxylating condensation generates chirality in a two-component. The activation parameters for the racemization of a series of ortho-alkyl-substituted diphenylacetic acids are determined.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsKinetic resolution of racemic 1-heteroarylalkanols by asymmetric esterification using diphenylacetic acid with pivalic anhydride and a chiral acyl-transfer catalyst is described. Molecular crystal of acridine and diphenylacetic acid and its absolute asymmetric photodecarboxylating condensation generates chirality in a two-component. The activation parameters for the racemization of a series of ortho-alkyl-substituted diphenylacetic acids are determined.
Solubility
Slightly soluble in water(0.13g/L).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
117-34-0 | |
212.248 | |
PYHXGXCGESYPCW-UHFFFAOYSA-N | |
8333 | |
2,2-diphenylacetic acid |
C14H12O2 | |
MFCD00004251 | |
diphenylacetic acid, diphenylethanoic acid, acetic acid, diphenyl, benzeneacetic acid, .alpha.-phenyl, 1,1-diphenylacetic acid, unii-658ncz0nko, diphenyl-acetic acid, alpha-toluic acid, alpha-phenyl, 658ncz0nko, benzeneacetic acid, alpha-phenyl | |
CHEBI:41967 | |
C1=CC=C(C=C1)C(C2=CC=CC=C2)C(=O)O |
Specifications
117-34-0 | |
195°C (25 mm) | |
C14H12O2 | |
100 g | |
14,3316 | |
Slightly soluble in water(0.13g/L). | |
C1=CC=C(C=C1)C(C2=CC=CC=C2)C(=O)O | |
212.248 | |
CHEBI:41967 | |
99% |
146°C to 148°C | |
99% | |
MFCD00004251 | |
1910978 | |
diphenylacetic acid, diphenylethanoic acid, acetic acid, diphenyl, benzeneacetic acid, .alpha.-phenyl, 1,1-diphenylacetic acid, unii-658ncz0nko, diphenyl-acetic acid, alpha-toluic acid, alpha-phenyl, 658ncz0nko, benzeneacetic acid, alpha-phenyl | |
PYHXGXCGESYPCW-UHFFFAOYSA-N | |
2,2-diphenylacetic acid | |
8333 | |
212.25 | |
Diphenylacetic Acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
EINECSNumber : 204-185-0
RTECSNumber : AH2515000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only