missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diphenylacetonitrile, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 150360025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Diphenylacetonitrile | |
73.0°C to 76.0°C | |
181.0°C (12.0 mmHg) | |
Authentic | |
Plastic bottle | |
(C6H5)2CHCN | |
09,674 | |
diphenylacetonitrile, dipan, diphenatrile, benzhydrylcyanide, diphenylmethylcyanide, benzyhydrylcyanide, acetonitrile, diphenyl, benzeneacetonitrile, alpha-phenyl, usaf kf-13, difenylacetonitril | |
NEBPTMCRLHKPOB-UHFFFAOYSA-N | |
2,2-diphenylacetonitrile | |
6837 | |
99+% |
86-29-3 | |
Cream to White or Yellow | |
120°C | |
99% min. (GC) | |
C14H11N | |
2.5 kg | |
15,3346 | |
Solubility in water: insoluble in water. Other solubilities: soluble in hot alcohol, propylene glycol, hot, ligroin and ether, slightly soluble in cold ligroin | |
C1=CC=C(C=C1)C(C#N)C2=CC=CC=C2 | |
193.25 | |
193.25 | |
Crystalline Powder |
Chemical Identifiers
86-29-3 | |
193.25 | |
diphenylacetonitrile, dipan, diphenatrile, benzhydrylcyanide, diphenylmethylcyanide, benzyhydrylcyanide, acetonitrile, diphenyl, benzeneacetonitrile, alpha-phenyl, usaf kf-13, difenylacetonitril | |
2,2-diphenylacetonitrile |
C14H11N | |
NEBPTMCRLHKPOB-UHFFFAOYSA-N | |
6837 | |
C1=CC=C(C=C1)C(C#N)C2=CC=CC=C2 |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 201-662-5
RUO – Research Use Only