Learn More
Diphenylacetonitrile, 99%, Thermo Scientific Chemicals
$61.00 - $202.71
Chemical Identifiers
CAS | 86-29-3 |
---|---|
Molecular Formula | C14H11N |
Molecular Weight (g/mol) | 193.249 |
MDL Number | MFCD00001862 |
InChI Key | NEBPTMCRLHKPOB-UHFFFAOYSA-N |
Synonym | diphenylacetonitrile, dipan, diphenatrile, benzhydrylcyanide, diphenylmethylcyanide, benzyhydrylcyanide, acetonitrile, diphenyl, benzeneacetonitrile, alpha-phenyl, usaf kf-13, difenylacetonitril |
PubChem CID | 6837 |
IUPAC Name | 2,2-diphenylacetonitrile |
SMILES | C1=CC=C(C=C1)C(C#N)C2=CC=CC=C2 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0240422
|
Thermo Scientific Chemicals
L0240422 |
100 g |
Each for $61.00
|
|
|||||
AAL0240436
|
Thermo Scientific Chemicals
L0240436 |
500 g |
Each for $202.71
|
|
|||||
Description
Diphenylacetonitrile undergoes anhydrous condensation with ethyl-4-bromo-butyrate. This is followed by its hydrolysis in alkaline medium that is used in the quantitative determination of 3-cyano-3,3-diphenylpropionic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiphenylacetonitrile undergoes anhydrous condensation with ethyl-4-bromo-butyrate. This is followed by its hydrolysis in alkaline medium that is used in the quantitative determination of 3-cyano-3,3-diphenylpropionic acid.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
86-29-3 | |
193.249 | |
NEBPTMCRLHKPOB-UHFFFAOYSA-N | |
6837 | |
C1=CC=C(C=C1)C(C#N)C2=CC=CC=C2 |
C14H11N | |
MFCD00001862 | |
diphenylacetonitrile, dipan, diphenatrile, benzhydrylcyanide, diphenylmethylcyanide, benzyhydrylcyanide, acetonitrile, diphenyl, benzeneacetonitrile, alpha-phenyl, usaf kf-13, difenylacetonitril | |
2,2-diphenylacetonitrile |
Specifications
86-29-3 | |
181°C (12 mmHg) | |
C14H11N | |
100 g | |
14,3316 | |
Insoluble in water. | |
C1=CC=C(C=C1)C(C#N)C2=CC=CC=C2 | |
193.249 | |
193.25 | |
Diphenylacetonitrile |
72°C to 76°C | |
120°C (248°F) | |
MFCD00001862 | |
1911160 | |
diphenylacetonitrile, dipan, diphenatrile, benzhydrylcyanide, diphenylmethylcyanide, benzyhydrylcyanide, acetonitrile, diphenyl, benzeneacetonitrile, alpha-phenyl, usaf kf-13, difenylacetonitril | |
NEBPTMCRLHKPOB-UHFFFAOYSA-N | |
2,2-diphenylacetonitrile | |
6837 | |
99% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H301-H315-H319-H335
EINECSNumber : 201-662-5
RTECSNumber : AL9800000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only