missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diphenylamine, 1% (w/v) in Sulfuric Acid, Ricca Chemical
$156.63 - $156.63
Chemical Identifiers
CAS | 122-39-4 |
---|---|
Molecular Formula | C12H11N |
Molecular Weight (g/mol) | 169.227 |
InChI Key | DMBHHRLKUKUOEG-UHFFFAOYSA-N |
Synonym | diphenylamine, n-phenylbenzenamine, anilinobenzene, benzenamine, n-phenyl, n,n-diphenylamine, phenylaniline, scaldip, n-phenylbenzeneamine, big dipper, no-scald |
PubChem CID | 11487 |
ChEBI | CHEBI:4640 |
IUPAC Name | N-phenylaniline |
SMILES | C1=CC=C(C=C1)NC2=CC=CC=C2 |
Chemical Identifiers
122-39-4 | |
169.227 | |
diphenylamine, n-phenylbenzenamine, anilinobenzene, benzenamine, n-phenyl, n,n-diphenylamine, phenylaniline, scaldip, n-phenylbenzeneamine, big dipper, no-scald | |
CHEBI:4640 | |
C1=CC=C(C=C1)NC2=CC=CC=C2 |
C12H11N | |
DMBHHRLKUKUOEG-UHFFFAOYSA-N | |
11487 | |
N-phenylaniline |
Specifications
122-39-4,7664-93-9 | |
100.0,0.55 | |
Amber Glass Bottle | |
120 mL | |
DMBHHRLKUKUOEG-UHFFFAOYSA-N | |
N-phenylaniline | |
11487 | |
1% (w/v) in Sulfuric Acid | |
Diphenylamine |
97.0,0.53 | |
Colorless | |
C12H11N | |
diphenylamine, n-phenylbenzenamine, anilinobenzene, benzenamine, n-phenyl, n,n-diphenylamine, phenylaniline, scaldip, n-phenylbenzeneamine, big dipper, no-scald | |
C1=CC=C(C=C1)NC2=CC=CC=C2 | |
169.227 | |
CHEBI:4640 | |
Liquid |
Safety and Handling
ShelfLife : 12 months