missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diphenylamine Hydrochloride 99.0+%, TCI America™

Supplier:  TCI America D087425G

Catalog No. D087425G


Explore available promotions

Description

Description

Specifications

Specifications

Diphenylamine Hydrochloride
180°C
C12H12ClN
25 g
[H+].[Cl-].N(C1=CC=CC=C1)C1=CC=CC=C1
205.69
205.69
Crystalline Powder
537-67-7
White
MFCD00042007
JEFJSEIUEJBMSR-UHFFFAOYSA-N
hydrogen N-phenylaniline chloride
68311
≥99.0% (HPLC,N)

Chemical Identifiers

537-67-7
205.69
JEFJSEIUEJBMSR-UHFFFAOYSA-N
hydrogen N-phenylaniline chloride
C12H12ClN
MFCD00042007
68311
[H+].[Cl-].N(C1=CC=CC=C1)C1=CC=CC=C1
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions