missing translation for 'onlineSavingsMsg'
Learn More

Diphosgene, 99%

CAS: 503-38-8 | C2Cl4O2 | 197.82 g/mol

$155.53 - $155.53

Chemical Identifiers

CAS 503-38-8
Molecular Formula C2Cl4O2
Molecular Weight (g/mol) 197.82
MDL Number MFCD00015553
InChI Key HCUYBXPSSCRKRF-UHFFFAOYSA-N
Synonym diphosgene, trichloromethyl chloroformate, diphosgen, difosgen czech, perchloromethyl formate, unii-po4q4r80lv, hsdb 371, carbonochloridic acid, trichloromethyl ester, methanol, trichloro-, chloroformate, chloroformic acid trichloromethyl ester
PubChem CID 10426
IUPAC Name trichloromethyl carbonochloridate
SMILES ClC(=O)OC(Cl)(Cl)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC243310100
SDS View Documents
Thermo Scientific Chemicals
243310100
10 mL Glass bottle
Each for $155.53
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
Specifications

Chemical Identifiers

503-38-8
197.82
HCUYBXPSSCRKRF-UHFFFAOYSA-N
10426
ClC(=O)OC(Cl)(Cl)Cl
C2Cl4O2
MFCD00015553
diphosgene, trichloromethyl chloroformate, diphosgen, difosgen czech, perchloromethyl formate, unii-po4q4r80lv, hsdb 371, carbonochloridic acid, trichloromethyl ester, methanol, trichloro-, chloroformate, chloroformic acid trichloromethyl ester
trichloromethyl carbonochloridate

Specifications

503-38-8
Colorless
128.0°C
98.5% min. (GC)
C2Cl4O2
ClCOOCCl3
10 mL
15, 3374
Solubility in water: decomposes. Other solubilities: soluble in alcohol, benzene and ether
ClC(=O)OC(Cl)(Cl)Cl
197.82
197.83
Liquid
-57.0°C
1.6400g/mL
Authentic
Glass bottle
1.4560 to 1.4590
MFCD00015553
1.64
diphosgene, trichloromethyl chloroformate, diphosgen, difosgen czech, perchloromethyl formate, unii-po4q4r80lv, hsdb 371, carbonochloridic acid, trichloromethyl ester, methanol, trichloro-, chloroformate, chloroformic acid trichloromethyl ester
HCUYBXPSSCRKRF-UHFFFAOYSA-N
trichloromethyl carbonochloridate
10426
99%
Diphosgene, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Fatal if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately

GHS Signal Word: Danger

EINECSNumber : 207-965-9

RTECSNumber : LQ7350000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only