Learn More
Diphosphoryl chloride, 90+%, Thermo Scientific™
Chemical Identifiers
CAS | 13498-14-1 |
---|---|
Molecular Formula | Cl4O3P2 |
Molecular Weight (g/mol) | 251.76 |
MDL Number | MFCD00061542 |
InChI Key | CNTIXUGILVWVHR-UHFFFAOYSA-N |
Synonym | diphosphoryl chloride, pyrophosphoryl chloride, diphosphoryl tetrachloride, diphosphoric tetrachloride, pyrophosphoric tetrachloride, dichlorophosphoryl oxyphosphonoyl dichloride, pyrophosphoryl tetrachloride, pyrophosphorylchloride, phosphorodichloridic anhydride |
PubChem CID | 4432411 |
SMILES | O=P(OP(=O)(Cl)Cl)(Cl)Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC393620050
|
Thermo Scientific Chemicals
393620050 |
5mL | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC393620250
|
Thermo Scientific Chemicals
393620250 |
25mL | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
13498-14-1 | |
251.76 | |
CNTIXUGILVWVHR-UHFFFAOYSA-N | |
4432411 |
Cl4O3P2 | |
MFCD00061542 | |
diphosphoryl chloride, pyrophosphoryl chloride, diphosphoryl tetrachloride, diphosphoric tetrachloride, pyrophosphoric tetrachloride, dichlorophosphoryl oxyphosphonoyl dichloride, pyrophosphoryl tetrachloride, pyrophosphorylchloride, phosphorodichloridic anhydride | |
O=P(OP(=O)(Cl)Cl)(Cl)Cl |
Specifications
13498-14-1 | |
90+% | |
MFCD00061542 | |
CNTIXUGILVWVHR-UHFFFAOYSA-N | |
251.76 | |
251.76 | |
Authentic | |
1.82 | |
Diphosphoryl chloride |
5mL | |
Cl4O3P2 | |
diphosphoryl chloride, pyrophosphoryl chloride, diphosphoryl tetrachloride, diphosphoric tetrachloride, pyrophosphoric tetrachloride, dichlorophosphoryl oxyphosphonoyl dichloride, pyrophosphoryl tetrachloride, pyrophosphorylchloride, phosphorodichloridic anhydride | |
O=P(OP(=O)(Cl)Cl)(Cl)Cl | |
4432411 | |
90+% | |
Glass bottle | |
1.8200g/mL |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 236-824-4
RUO â Research Use Only