missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dipropyl phthalate, 99+%, Thermo Scientific Chemicals

$167.45 - $432.44

Chemical Identifiers

CAS 131-16-8
InChI Key MQHNKCZKNAJROC-UHFFFAOYSA-N
Synonym dipropyl phthalate, di-n-propyl phthalate, di-n-propylphthalate, phthalic acid dipropyl ester, phthalic acid, dipropyl ester, phthalic acid di-n-propyl ester, 1,2-benzenedicarboxylic acid, dipropyl ester, dipropylphthalate, n-dipropyl phthalate, unii-8usp5y77ss
PubChem CID 8559
ChEBI CHEBI:60069
IUPAC Name dipropyl benzene-1,2-dicarboxylate
SMILES CCCOC(=O)C1=CC=CC=C1C(=O)OCCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC409160250
SDS View Documents
Thermo Scientific Chemicals
409160250
25 g Glass bottle
Each for $167.45
 
AC409161000
SDS View Documents
Thermo Scientific Chemicals
409161000
100 g Glass bottle
Each for $432.44
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

131-16-8
dipropyl phthalate, di-n-propyl phthalate, di-n-propylphthalate, phthalic acid dipropyl ester, phthalic acid, dipropyl ester, phthalic acid di-n-propyl ester, 1,2-benzenedicarboxylic acid, dipropyl ester, dipropylphthalate, n-dipropyl phthalate, unii-8usp5y77ss
CHEBI:60069
CCCOC(=O)C1=CC=CC=C1C(=O)OCCC
MQHNKCZKNAJROC-UHFFFAOYSA-N
8559
dipropyl benzene-1,2-dicarboxylate

Specifications

131-16-8
1.0780g/mL
>110°C
99+%
C14H18O4
25 g
dipropyl phthalate, di-n-propyl phthalate, di-n-propylphthalate, phthalic acid dipropyl ester, phthalic acid, dipropyl ester, phthalic acid di-n-propyl ester, 1,2-benzenedicarboxylic acid, dipropyl ester, dipropylphthalate, n-dipropyl phthalate, unii-8usp5y77ss
MQHNKCZKNAJROC-UHFFFAOYSA-N
dipropyl benzene-1,2-dicarboxylate
8559
250.29
Liquid
Colorless
317.5°C
Authentic
Glass bottle
1.4960 to 1.4980
1.078
Solubility in water: in water: 0.015% by weight.
CCCOC(=O)C1=CC=CC=C1C(=O)OCCC
250.29
CHEBI:60069
99+%
Dipropyl phthalate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Suspected of causing cancer.

GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 205-015-8

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only