missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dipropylene glycol, mixture of isomers, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1871636

 View more versions of this product

Catalog No. AAA1871636


Explore available promotions

Description

Description

Dipropylene glycol, mixture of isomers is used as a solvent, plasticizer, polymerization initiator and monomer. It acts as an additive used in perfumes, skin and hair care products. It is an active ingredient in commercial fog fluid, which is used as an entertainment in fog machines. Further, it is used as an intermediate in polyurethane polyols, alkyd resins and non-reinforced polyesters. In addition to this, it is used in dyes, paints, hydraulic brake fluids and cutting oils.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dipropylene glycol, mixture of isomers is used as a solvent, plasticizer, polymerization initiator and monomer. It acts as an additive used in perfumes, skin and hair care products. It is an active ingredient in commercial fog fluid, which is used as an entertainment in fog machines. Further, it is used as an intermediate in polyurethane polyols, alkyd resins and non-reinforced polyesters. In addition to this, it is used in dyes, paints, hydraulic brake fluids and cutting oils.

Solubility
Miscible with cold water, hot water, methanol, diethyl ether and ethanol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Specifications

Specifications

Dipropylene glycol
25265-71-8
1.023
0.99
1.441
500 g
Miscible with cold water,hot water,methanol,diethyl ether and ethanol.
CC(O)COCC(C)O
134.18
134.18
mixture of isomers
°C to 32°C
231°C to 235°C
C6H14O3
MFCD00004538
1,1'-oxydi-2-propanol, 2-propanol, 1,1'-oxybis, 1,1'-oxydipropan-2-ol, bis 2-hydroxypropyl ether, 4-oxaheptane-2,6-diol, 1,1'-oxybis-2-propanol, 2,2'-dihydroxydipropyl ether, 2-propanol, 1,1'-oxydi, niax catalyst d-19, 1,1'-dimethyldiethylene glycol
AZUXKVXMJOIAOF-UHFFFAOYNA-N
1-(2-hydroxypropoxy)propan-2-ol
8087

Chemical Identifiers

25265-71-8
AZUXKVXMJOIAOF-UHFFFAOYNA-N
1-(2-hydroxypropoxy)propan-2-ol
134.18
8087
CC(O)COCC(C)O
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 246-770-3

RTECSNumber : UB9765000

TSCA : Yes

storageNote1 : Hygroscopic

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only