missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Direct Red 80

CAS: 2610-10-8 | C45H26N10Na6O21S6 | 1373.05 g/mol

$75.55 - $508.41

Chemical Identifiers

CAS 2610-10-8
MDL Number MFCD00054389
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2169306
View Documents
Thermo Scientific Chemicals
B2169306
5 g N/A
Only null left
 
AAB2169314
View Documents
Thermo Scientific Chemicals
B2169314
25 g N/A
Only null left
 
AAB2169322
View Documents
Thermo Scientific Chemicals
B2169322
100 g N/A
Only null left
 
Description

Description

Direct red 80 is a dye useful as a cardiac muscle stain and as a model compound in adsorption studies. It has also been utilized to compare efficacy of textile dye removal in aqueous solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Direct red 80 is a dye useful as a cardiac muscle stain and as a model compound in adsorption studies. It has also been utilized to compare efficacy of textile dye removal in aqueous solutions.

Solubility
Partly soluble in water, very slightly soluble in ethanol, and cellosolve. Insoluble in many organic solvents.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

2610-10-8
MFCD00054389

Specifications

2610-10-8
MFCD00054389
LDCKXVAPQFUIOI-ZFLDVXHKSA-H
hexasodium;4-oxo-7-[[5-oxo-7-sulfonato-6-[[2-sulfonato-4-[(4-sulfonatophenyl)diazenyl]phenyl]hydrazinylidene]naphthalen-2-yl]carbamoylamino]-3-[[2-sulfonato-4-[(4-sulfonatophenyl)diazenyl]phenyl]hydrazinylidene]naphthalene-2-sulfonate
57369623
5 g
C45H26N10Na6O21S6
Partly soluble in water,very slightly soluble in ethanol,and cellosolve. Insoluble in many organic solvents.
C1=CC(=CC=C1N=NC2=CC(=C(C=C2)NN=C3C(=CC4=C(C3=O)C=CC(=C4)NC(=O)NC5=CC6=C(C=C5)C(=O)C(=NNC7=C(C=C(C=C7)N=NC8=CC=C(C=C8)S(=O)(=O)[O-])S(=O)(=O)[O-])C(=C6)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+]
1373.05
1373.09
Direct Red 80
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 220-027-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only