BrandTech™ Discharge Tubes for BRAND Dispensette S Bottletop Dispensers with standard valves
Discharge tubes to fit BRAND Dispensettes made 2016 and later
Supplier:  BrandTech™ 708019
 View more versions of this product
Catalog No. 14-380-896
Explore available promotions
Description
Description
  • Discharge tubes come in two sizes: For instruments with nominal capacity 1 mL, 2 mL, 5 mL, and 10 mL; and for instruments with nominal capacity of 25 mL, 50 mL and 100 mL
  • Choose from standard and fine tip models
  • Dispensette S dispensers have red trim
  • Dispensette S Organic dispensers have yellow trim
Specifications
Specifications
Discharge Tube
White/Yellow
Dispensing liquid
Discharge Tube, Safety Cap
Autoclavable
ETFE, PTFE, Polypropylene
Dispensette™ S Organic, nominal volumes 25, 50 and 10 mL