missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Disodium alpha-Glycerophosphate Hydrate 85.0+%, TCI America™
Supplier: TCI America G009625G
Specifications
Disodium alpha-Glycerophosphate Hydrate | |
White | |
MFCD00035539 | |
sodium glycerophosphate, disodium alpha-glycerophosphate, alpha-glycerophosphoric acid disodium salt, 1,2,3-propanetriol, mono dihydrogen phosphate , disodium salt, 1,2,3-propanetriol, 1-dihydrogen phosphate , disodium salt, dipotassium glyceryl phosphate, 1,2,3-propanetriol, 1-dihydrogen phosphate , sodium salt 1:2, 1,2,3-propanetriol, mono dihydrogen phosphate , sodium salt 1:2, 57-03-4 parent, sodium 1-glycerophosphate | |
C(C(COP(=O)([O-])[O-])O)O.[Na+].[Na+] | |
216.036 | |
216.04 | |
Crystalline Powder |
1555-56-2 | |
C3H7Na2O6P | |
25 g | |
GEKBIENFFVFKRG-UHFFFAOYSA-L | |
disodium;2,3-dihydroxypropyl phosphate | |
14754 | |
≥85.0% (T) |
Chemical Identifiers
1555-56-2 | |
216.036 | |
GEKBIENFFVFKRG-UHFFFAOYSA-L | |
14754 | |
C(C(COP(=O)([O-])[O-])O)O.[Na+].[Na+] |
C3H7Na2O6P | |
MFCD00035539 | |
sodium glycerophosphate, disodium alpha-glycerophosphate, alpha-glycerophosphoric acid disodium salt, 1,2,3-propanetriol, mono dihydrogen phosphate , disodium salt, 1,2,3-propanetriol, 1-dihydrogen phosphate , disodium salt, dipotassium glyceryl phosphate, 1,2,3-propanetriol, 1-dihydrogen phosphate , sodium salt 1:2, 1,2,3-propanetriol, mono dihydrogen phosphate , sodium salt 1:2, 57-03-4 parent, sodium 1-glycerophosphate | |
disodium;2,3-dihydroxypropyl phosphate |
Safety and Handling
EINECSNumber : (2)-1905&(2)-1990
TSCA : Yes