missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Disodium terephthalate, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 04294614

 View more versions of this product

Catalog No. AA4294614

Explore available promotions

Description

Description

Disodium terephthalate and its various derivatives are synthesized via simple acid-base chemistry for anode materials in Na ion batteries. They show excellent electrochemical performance, including little capacity fading over 90 cycles, ideal redox potential, and excellent rate performance, making them promising candidates for Na ion batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Disodium terephthalate
C8H4Na2O4
25 g
Soluble in water.
C1=CC(=CC=C1C(=O)[O-])C(=O)[O-].[Na+].[Na+]
210.096
210.1
Powder
10028-70-3
MFCD00013137
disodium terephthalate, terephthalic acid disodium salt, terephthalic acid, disodium salt, sodium terephthalate, unii-gi30zoc0oo, gi30zoc0oo, disodiumterephthalate, acmc-1bvw0, disodium terephthalate 5g, 1,4-benzenedicarboxylicacid, sodium salt 1:?
VIQSRHWJEKERKR-UHFFFAOYSA-L
disodium;terephthalate
82305
≥99%

Chemical Identifiers

10028-70-3
210.096
VIQSRHWJEKERKR-UHFFFAOYSA-L
82305
C1=CC(=CC=C1C(=O)[O-])C(=O)[O-].[Na+].[Na+]
C8H4Na2O4
MFCD00013137
disodium terephthalate, terephthalic acid disodium salt, terephthalic acid, disodium salt, sodium terephthalate, unii-gi30zoc0oo, gi30zoc0oo, disodiumterephthalate, acmc-1bvw0, disodium terephthalate 5g, 1,4-benzenedicarboxylicacid, sodium salt 1:?
disodium;terephthalate
Safety and Handling

Safety and Handling

EINECSNumber : 233-075-5

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only