missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BrandTech™ BRAND™ Dispensette™ S Digital Bottletop Dispensers

The Dispensette™ S Digital Bottletop Dispensers feature a floating piston design for low-force operation and no wearing parts. Optimized flow channels make dispensing and priming even easier-particularly in instruments with large volumes. Designed for general-purpose dispensing applications.

Supplier:  BrandTech™ BRAND™ 4600310

 View more versions of this product

Catalog No. 13-689-006


Explore available promotions

Includes: Discharge tube, valve mounting/calibration tool, polypropylene adapters, telescoping filling tube, recirculation tube (optional), performance certificate, operating manual

Description

Description

The Dispensette™ S Digital Bottletop Dispensers feature a floating piston design for low-force operation and no wearing parts. Optimized flow channels make dispensing and priming even easier-particularly in instruments with large volumes. Designed for g

  • New discharge tube, with or without recirculation valve
  • New valve system - no additional sealing rings required, facilitates cleaning and preparations for autoclaving
  • Simple priming by optimized flow channels
  • Less dispensing force - especially with large volume instruments
  • Easy Calibration technique for calibration adjustment in seconds without tools
  • Pivoted screw cap - does not disturb dispensing
  • Easy to calibrate and adjust in order to comply with GLP guidelines
  • Easy to dismantle for cleaning
  • Replacable filling and discharge valve with safety bulb (locks when discharge tube is not assembled)
  • 360° rotatable valve block allows bottle label to face user
  • Telescoping filling tube adjusts easily to different size bottles
  • DE-M marking

Warranty

One year

Specifications

Specifications

0.6% (6μL)
Autoclavable (121°C)
Red
0.1 to 1 mL
Dispensing aggressive reagents, including concentrated bases and acids such as H3PO4, H2SO4 (except e.g., HCl, HNO3 und HF), saline solutions, as well as many organic solvents
Dispenser
Certifications: ISO 9001
Without recirculation valve
Digital
0.005 mL
Videos
SDS
Documents

Documents

Promotions

Promotions