Fisherbrand™ Face Shield
High-quality, optically clear disposable face shield
Supplier:  Fisherbrand™ 4505AF
Catalog No. 19-181-800C
Explore available promotions
Description
Description
Fisherbrand™ Face Shields are lightweight and comfortable. They provide protection from splashing liquids, blood, and other bodily fluids and are ideal for use in laboratories, medical settings, dentistry, and veterinary medicine
  • Medical face shield
  • Optical grade PET construction
  • Anti-fog and non-glare design
  • Flexible construction with comfortable polyurethane foam headband
  • Secures to head with elastic band
  • Latex free
  • FDA Class 2 Medical Device, 510(k)
  • Not approved for medical use in Canada
Specifications
Specifications
FDA Class 2 Medical Device, 510(k)
Blue, Clear
Polyurethane Foam
144/Cs.
Single-use
Full Faceshield, 7.5 in. Length
Disposable Faceshield
7.5 in.
One Size