missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Disposable Frocks
White 58 gsm polypropylene and polyethylene frocks that create a barrier against particles, micro-organisms and nonhazardous liquids.
$115.38 - $137.50
Specifications
Product Type | Knee-length Frock |
---|---|
Material | Polypropylene, Polyethylene |
Color | White |
Disposable | Single-use |
Sterility | Non-sterile |
Description
- Front zipper closure
- Elastic cuffs
- Snap-closure collar
- Knee-length coverage
Specifications
Knee-length Frock | |
White | |
Non-sterile | |
Elastic |
Polypropylene, Polyethylene | |
Single-use | |
Zipper | |
58 gsm |